Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33029798-33030071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1
Entrez Gene ID
3301
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
Gene Aliases
DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2
UniGene
Hs.445203
Gene Chromosome Location
Chr.9: 33025203-33039907 on Build GRCh38
Transcripts
NM_001314039.1 NM_001539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6367436 c.365C>A p.A122E Chr.9: 33029939-33029939 on Build GRCh38
6647039 c.416+8delT p.? Chr.9: 33029998-33029998 on Build GRCh38
3906868 c.409T>C p.C137R Chr.9: 33029983-33029983 on Build GRCh38
6412427 c.360T>A p.N120K Chr.9: 33029934-33029934 on Build GRCh38
6387932 c.325C>T p.H109Y Chr.9: 33029899-33029899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916457048 Chr.9: 33029930-33029930 on Build GRCh38
rs778896041 Chr.9: 33030020-33030020 on Build GRCh38
rs757739036 Chr.9: 33029989-33029989 on Build GRCh38
rs375525655 Chr.9: 33029993-33029993 on Build GRCh38
rs766344691 Chr.9: 33030003-33030010 on Build GRCh38
rs552651063 Chr.9: 33030001-33030005 on Build GRCh38
rs772024356 Chr.9: 33030035-33030035 on Build GRCh38
rs926467596 Chr.9: 33029831-33029831 on Build GRCh38
rs768144940 Chr.9: 33029863-33029869 on Build GRCh38
rs567634635 Chr.9: 33030044-33030044 on Build GRCh38
rs764787440 Chr.9: 33029949-33029949 on Build GRCh38
rs780249843 Chr.9: 33030002-33030002 on Build GRCh38
rs747769515 Chr.9: 33029997-33029998 on Build GRCh38
rs199981412 Chr.9: 33030011-33030011 on Build GRCh38
rs528687736 Chr.9: 33029841-33029841 on Build GRCh38
rs991549582 Chr.9: 33029920-33029920 on Build GRCh38
rs760438476 Chr.9: 33029917-33029917 on Build GRCh38
rs762223116 Chr.9: 33030041-33030041 on Build GRCh38
rs927811037 Chr.9: 33030021-33030021 on Build GRCh38
rs768789351 Chr.9: 33029873-33029873 on Build GRCh38
rs773064898 Chr.9: 33030040-33030040 on Build GRCh38
rs200841542 Chr.9: 33029916-33029916 on Build GRCh38
rs914592363 Chr.9: 33029964-33029964 on Build GRCh38
rs768879330 Chr.9: 33030014-33030014 on Build GRCh38
rs982315131 Chr.9: 33030003-33030003 on Build GRCh38
rs878896678 Chr.9: 33029925-33029925 on Build GRCh38
rs774061267 Chr.9: 33029879-33029879 on Build GRCh38
rs552013421 Chr.9: 33029892-33029892 on Build GRCh38
rs903485051 Chr.9: 33029998-33030002 on Build GRCh38
rs369423129 Chr.9: 33029994-33029994 on Build GRCh38
rs765592173 Chr.9: 33030042-33030042 on Build GRCh38
rs770004997 Chr.9: 33029835-33029835 on Build GRCh38
rs760348780 Chr.9: 33030001-33030010 on Build GRCh38
rs372705804 Chr.9: 33029929-33029929 on Build GRCh38
rs759893203 Chr.9: 33029953-33029953 on Build GRCh38
rs531258329 Chr.9: 33029941-33029941 on Build GRCh38
rs774257322 Chr.9: 33029874-33029874 on Build GRCh38
rs748199635 Chr.9: 33030023-33030023 on Build GRCh38
rs567434020 Chr.9: 33030000-33030001 on Build GRCh38
rs780186510 Chr.9: 33029838-33029838 on Build GRCh38
rs771935926 Chr.9: 33029895-33029895 on Build GRCh38

View Full Product Details