Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2029466-2029699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRD
Entrez Gene ID
2563
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Gene Aliases
EIG10, EJM7, GEFSP5
UniGene
Hs.113882
Gene Chromosome Location
Chr.1: 2019329-2030753 on Build GRCh38
Transcripts
NM_000815.4 XM_011541194.2 XM_017000936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668638 c.910G>A p.A304T Chr.1: 2029613-2029613 on Build GRCh38
463652 c.968C>T p.A323V Chr.1: 2029671-2029671 on Build GRCh38
2124564 c.960C>T p.F320F Chr.1: 2029663-2029663 on Build GRCh38
404646 c.934G>A p.V312I Chr.1: 2029637-2029637 on Build GRCh38
7297379 c.982T>C p.Y328H Chr.1: 2029685-2029685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754466489 Chr.1: 2029560-2029560 on Build GRCh38
rs139676179 Chr.1: 2029645-2029645 on Build GRCh38
rs116197762 Chr.1: 2029525-2029525 on Build GRCh38
rs985336803 Chr.1: 2029512-2029512 on Build GRCh38
rs760757497 Chr.1: 2029570-2029572 on Build GRCh38
rs200124298 Chr.1: 2029636-2029636 on Build GRCh38
rs896016478 Chr.1: 2029673-2029673 on Build GRCh38
rs1026792771 Chr.1: 2029489-2029489 on Build GRCh38
rs757483435 Chr.1: 2029588-2029588 on Build GRCh38
rs755449066 Chr.1: 2029505-2029505 on Build GRCh38
rs747305505 Chr.1: 2029579-2029579 on Build GRCh38
rs377674028 Chr.1: 2029523-2029523 on Build GRCh38
rs772061844 Chr.1: 2029653-2029653 on Build GRCh38
rs375250289 Chr.1: 2029513-2029513 on Build GRCh38
rs921986335 Chr.1: 2029543-2029543 on Build GRCh38
rs376640236 Chr.1: 2029666-2029666 on Build GRCh38
rs781426402 Chr.1: 2029611-2029611 on Build GRCh38
rs200391871 Chr.1: 2029595-2029595 on Build GRCh38
rs28581504 Chr.1: 2029487-2029487 on Build GRCh38
rs937477150 Chr.1: 2029669-2029669 on Build GRCh38
rs568302498 Chr.1: 2029534-2029534 on Build GRCh38
rs146381127 Chr.1: 2029672-2029672 on Build GRCh38
rs372178849 Chr.1: 2029561-2029561 on Build GRCh38
rs768500052 Chr.1: 2029531-2029531 on Build GRCh38
rs774201718 Chr.1: 2029637-2029637 on Build GRCh38
rs370906086 Chr.1: 2029524-2029524 on Build GRCh38
rs948197538 Chr.1: 2029649-2029649 on Build GRCh38
rs750156130 Chr.1: 2029537-2029537 on Build GRCh38
rs201101812 Chr.1: 2029663-2029663 on Build GRCh38
rs370920369 Chr.1: 2029504-2029504 on Build GRCh38
rs548401742 Chr.1: 2029528-2029528 on Build GRCh38
rs748483885 Chr.1: 2029510-2029510 on Build GRCh38
rs774300906 Chr.1: 2029532-2029532 on Build GRCh38
rs951319501 Chr.1: 2029491-2029491 on Build GRCh38
rs753305124 Chr.1: 2029555-2029555 on Build GRCh38
rs368765147 Chr.1: 2029675-2029675 on Build GRCh38
rs755731843 Chr.1: 2029503-2029503 on Build GRCh38
rs765992334 Chr.1: 2029546-2029546 on Build GRCh38
rs778104822 Chr.1: 2029578-2029578 on Build GRCh38
rs368572778 Chr.1: 2029545-2029545 on Build GRCh38
rs150619024 Chr.1: 2029570-2029570 on Build GRCh38
rs866215877 Chr.1: 2029613-2029613 on Build GRCh38
rs373955279 Chr.1: 2029507-2029507 on Build GRCh38
rs745961044 Chr.1: 2029631-2029631 on Build GRCh38

View Full Product Details