Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127668969-127669230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTU
Entrez Gene ID
27152
Gene Name
inturned planar cell polarity protein
Gene Aliases
INT, PDZD6, PDZK6
UniGene
Hs.391481
Gene Chromosome Location
Chr.4: 127623271-127716779 on Build GRCh38
Transcripts
NM_015693.3 XM_011531844.2 XM_011531845.2 XM_011531846.1 XM_011531847.1 XM_011531849.2 XM_011531850.2 XM_011531851.2 XM_017008025.1 XM_017008026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941668 c.1084G>A p.V362I Chr.4: 127669147-127669147 on Build GRCh38
393648 c.973-2A>G p.? Chr.4: 127669034-127669034 on Build GRCh38
1618366 c.1063G>T p.E355* Chr.4: 127669126-127669126 on Build GRCh38
3660879 c.1063G>T p.E355* Chr.4: 127669126-127669126 on Build GRCh38
1051072 c.1068C>T p.N356N Chr.4: 127669131-127669131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139082309 Chr.4: 127669090-127669090 on Build GRCh38
rs769052353 Chr.4: 127669131-127669131 on Build GRCh38
rs942171585 Chr.4: 127669120-127669120 on Build GRCh38
rs780059759 Chr.4: 127669100-127669100 on Build GRCh38
rs372532881 Chr.4: 127669151-127669151 on Build GRCh38
rs779685404 Chr.4: 127669114-127669115 on Build GRCh38
rs774745487 Chr.4: 127669175-127669175 on Build GRCh38
rs745827606 Chr.4: 127669097-127669097 on Build GRCh38
rs751028154 Chr.4: 127669168-127669168 on Build GRCh38
rs776885562 Chr.4: 127669132-127669132 on Build GRCh38
rs192164921 Chr.4: 127669064-127669064 on Build GRCh38
rs149126116 Chr.4: 127669045-127669045 on Build GRCh38
rs755820204 Chr.4: 127669053-127669053 on Build GRCh38
rs994140686 Chr.4: 127669143-127669143 on Build GRCh38
rs758341002 Chr.4: 127669098-127669098 on Build GRCh38
rs911379401 Chr.4: 127669108-127669108 on Build GRCh38
rs143236402 Chr.4: 127669047-127669047 on Build GRCh38
rs556155950 Chr.4: 127669086-127669086 on Build GRCh38
rs766856591 Chr.4: 127669173-127669173 on Build GRCh38
rs773109775 Chr.4: 127669161-127669161 on Build GRCh38
rs748279179 Chr.4: 127669141-127669141 on Build GRCh38
rs754467084 Chr.4: 127669183-127669184 on Build GRCh38
rs74722824 Chr.4: 127669117-127669117 on Build GRCh38
rs1037828930 Chr.4: 127669126-127669126 on Build GRCh38
rs753327709 Chr.4: 127669084-127669084 on Build GRCh38
rs763522882 Chr.4: 127669172-127669172 on Build GRCh38

View Full Product Details