Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67078393-67078618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELZ
Entrez Gene ID
9931
Gene Name
helicase with zinc finger
Gene Aliases
DHRC, DRHC, HUMORF5
UniGene
Hs.370140
Gene Chromosome Location
Chr.17: 67070438-67245989 on Build GRCh38
Transcripts
NM_014877.3 XM_005257888.4 XM_005257889.4 XM_006722214.3 XM_006722215.3 XM_006722216.3 XM_011525544.2 XM_017025477.1 XM_017025478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2797322 c.5675C>T p.P1892L Chr.17: 67078406-67078406 on Build GRCh38
7209536 c.5497C>A p.P1833T Chr.17: 67078584-67078584 on Build GRCh38
4951897 c.5626G>A p.A1876T Chr.17: 67078455-67078455 on Build GRCh38
4689987 c.5648G>T p.S1883I Chr.17: 67078433-67078433 on Build GRCh38
2797325 c.5515C>A p.P1839T Chr.17: 67078566-67078566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766516759 Chr.17: 67078413-67078413 on Build GRCh38
rs374888335 Chr.17: 67078534-67078534 on Build GRCh38
rs769194308 Chr.17: 67078465-67078465 on Build GRCh38
rs758289523 Chr.17: 67078411-67078412 on Build GRCh38
rs866515391 Chr.17: 67078426-67078426 on Build GRCh38
rs201001169 Chr.17: 67078586-67078586 on Build GRCh38
rs536897964 Chr.17: 67078543-67078543 on Build GRCh38
rs749907469 Chr.17: 67078509-67078509 on Build GRCh38
rs199927742 Chr.17: 67078568-67078568 on Build GRCh38
rs778786355 Chr.17: 67078447-67078447 on Build GRCh38
rs762308723 Chr.17: 67078504-67078504 on Build GRCh38
rs980491145 Chr.17: 67078430-67078430 on Build GRCh38
rs774546962 Chr.17: 67078533-67078533 on Build GRCh38
rs763280305 Chr.17: 67078412-67078412 on Build GRCh38
rs745477812 Chr.17: 67078453-67078453 on Build GRCh38
rs754379098 Chr.17: 67078591-67078591 on Build GRCh38
rs763225092 Chr.17: 67078481-67078481 on Build GRCh38
rs201422510 Chr.17: 67078418-67078418 on Build GRCh38
rs373138572 Chr.17: 67078477-67078477 on Build GRCh38
rs753664258 Chr.17: 67078544-67078544 on Build GRCh38
rs908624633 Chr.17: 67078576-67078576 on Build GRCh38
rs200149411 Chr.17: 67078585-67078585 on Build GRCh38
rs566283236 Chr.17: 67078456-67078456 on Build GRCh38
rs769994003 Chr.17: 67078478-67078478 on Build GRCh38
rs200688677 Chr.17: 67078580-67078580 on Build GRCh38
rs181902989 Chr.17: 67078417-67078417 on Build GRCh38
rs751718696 Chr.17: 67078414-67078414 on Build GRCh38
rs746518693 Chr.17: 67078589-67078589 on Build GRCh38
rs372477799 Chr.17: 67078578-67078578 on Build GRCh38
rs773733173 Chr.17: 67078480-67078480 on Build GRCh38
rs866349696 Chr.17: 67078446-67078446 on Build GRCh38
rs758080750 Chr.17: 67078454-67078454 on Build GRCh38
rs756939700 Chr.17: 67078442-67078442 on Build GRCh38
rs369464897 Chr.17: 67078455-67078455 on Build GRCh38
rs766745743 Chr.17: 67078524-67078524 on Build GRCh38

View Full Product Details