Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102349887-102350138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC2
Entrez Gene ID
329
Gene Name
baculoviral IAP repeat containing 2
Gene Aliases
API1, HIAP2, Hiap-2, MIHB, RNF48, c-IAP1, cIAP1
UniGene
Hs.696238
Gene Chromosome Location
Chr.11: 102347182-102378670 on Build GRCh38
Transcripts
NM_001166.4 NM_001256163.1 NM_001256166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291874 c.36T>C p.G12G Chr.11: 102349890-102349890 on Build GRCh38
4849421 c.155C>T p.T52I Chr.11: 102350009-102350009 on Build GRCh38
378986 c.171C>T p.P57P Chr.11: 102350025-102350025 on Build GRCh38
159372 c.91T>A p.W31R Chr.11: 102349945-102349945 on Build GRCh38
7400273 c.78G>A p.T26T Chr.11: 102349932-102349932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899855245 Chr.11: 102349964-102349964 on Build GRCh38
rs756727284 Chr.11: 102350040-102350040 on Build GRCh38
rs745504549 Chr.11: 102349983-102349983 on Build GRCh38
rs765313317 Chr.11: 102349934-102349934 on Build GRCh38
rs750592727 Chr.11: 102349937-102349937 on Build GRCh38
rs889143031 Chr.11: 102349948-102349948 on Build GRCh38
rs745428800 Chr.11: 102350059-102350059 on Build GRCh38
rs771672336 Chr.11: 102349986-102349986 on Build GRCh38
rs190798003 Chr.11: 102350026-102350026 on Build GRCh38
rs368571070 Chr.11: 102349955-102349955 on Build GRCh38
rs760639875 Chr.11: 102349929-102349929 on Build GRCh38
rs763995359 Chr.11: 102349930-102349930 on Build GRCh38
rs756781770 Chr.11: 102349971-102349971 on Build GRCh38
rs549630656 Chr.11: 102350108-102350108 on Build GRCh38
rs369289038 Chr.11: 102349963-102349963 on Build GRCh38
rs143071976 Chr.11: 102349958-102349958 on Build GRCh38
rs773323674 Chr.11: 102350109-102350109 on Build GRCh38
rs373468213 Chr.11: 102350083-102350083 on Build GRCh38
rs561590437 Chr.11: 102349953-102349953 on Build GRCh38
rs753218434 Chr.11: 102350037-102350037 on Build GRCh38
rs781738757 Chr.11: 102349962-102349962 on Build GRCh38
rs755242949 Chr.11: 102350031-102350031 on Build GRCh38
rs147638046 Chr.11: 102350017-102350017 on Build GRCh38
rs776687396 Chr.11: 102349931-102349931 on Build GRCh38
rs775343215 Chr.11: 102349992-102349992 on Build GRCh38
rs778599181 Chr.11: 102349974-102349974 on Build GRCh38
rs570604037 Chr.11: 102350028-102350028 on Build GRCh38
rs765386384 Chr.11: 102350018-102350018 on Build GRCh38
rs776440324 Chr.11: 102350106-102350106 on Build GRCh38
rs1028016459 Chr.11: 102350016-102350016 on Build GRCh38
rs991894330 Chr.11: 102350100-102350100 on Build GRCh38
rs768018809 Chr.11: 102350032-102350032 on Build GRCh38
rs763208707 Chr.11: 102350025-102350025 on Build GRCh38
rs200457538 Chr.11: 102350056-102350056 on Build GRCh38
rs539386469 Chr.11: 102350052-102350052 on Build GRCh38
rs201622293 Chr.11: 102350034-102350034 on Build GRCh38
rs746846483 Chr.11: 102350013-102350013 on Build GRCh38
rs548310302 Chr.11: 102349932-102349932 on Build GRCh38
rs768443468 Chr.11: 102350014-102350014 on Build GRCh38
rs766495671 Chr.11: 102350022-102350022 on Build GRCh38
rs375296936 Chr.11: 102350099-102350099 on Build GRCh38

View Full Product Details