Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44448049-44448308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD8
Entrez Gene ID
386617
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
UniGene
Hs.479644
Gene Chromosome Location
Chr.4: 44173903-44448885 on Build GRCh38
Transcripts
NM_198353.2 XM_011513690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692131 c.343G>A p.D115N Chr.4: 44448181-44448181 on Build GRCh38
1670981 c.364G>T p.E122* Chr.4: 44448160-44448160 on Build GRCh38
6692127 c.220T>C p.F74L Chr.4: 44448304-44448304 on Build GRCh38
4694830 c.441G>A p.S147S Chr.4: 44448083-44448083 on Build GRCh38
6167598 c.401C>A p.A134D Chr.4: 44448123-44448123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032206755 Chr.4: 44448102-44448102 on Build GRCh38
rs751619148 Chr.4: 44448179-44448179 on Build GRCh38
rs372956006 Chr.4: 44448286-44448286 on Build GRCh38
rs777815401 Chr.4: 44448138-44448138 on Build GRCh38
rs140959192 Chr.4: 44448069-44448069 on Build GRCh38
rs768530413 Chr.4: 44448159-44448159 on Build GRCh38
rs924945852 Chr.4: 44448084-44448084 on Build GRCh38
rs748990834 Chr.4: 44448156-44448156 on Build GRCh38
rs776941842 Chr.4: 44448259-44448259 on Build GRCh38
rs199599084 Chr.4: 44448168-44448168 on Build GRCh38
rs1008308742 Chr.4: 44448178-44448178 on Build GRCh38
rs778840116 Chr.4: 44448221-44448221 on Build GRCh38
rs770479018 Chr.4: 44448253-44448253 on Build GRCh38
rs777588352 Chr.4: 44448243-44448243 on Build GRCh38
rs368933267 Chr.4: 44448247-44448247 on Build GRCh38
rs752917520 Chr.4: 44448122-44448122 on Build GRCh38
rs939088004 Chr.4: 44448281-44448281 on Build GRCh38
rs551410926 Chr.4: 44448268-44448268 on Build GRCh38
rs767141209 Chr.4: 44448073-44448073 on Build GRCh38
rs377247366 Chr.4: 44448129-44448129 on Build GRCh38
rs778642107 Chr.4: 44448160-44448160 on Build GRCh38
rs779819635 Chr.4: 44448197-44448197 on Build GRCh38
rs757384749 Chr.4: 44448194-44448194 on Build GRCh38
rs780090301 Chr.4: 44448155-44448155 on Build GRCh38
rs956366535 Chr.4: 44448103-44448103 on Build GRCh38
rs751842712 Chr.4: 44448273-44448273 on Build GRCh38
rs755392751 Chr.4: 44448108-44448108 on Build GRCh38
rs767649000 Chr.4: 44448278-44448278 on Build GRCh38
rs750526750 Chr.4: 44448284-44448284 on Build GRCh38
rs753819539 Chr.4: 44448200-44448200 on Build GRCh38
rs764343485 Chr.4: 44448177-44448177 on Build GRCh38
rs961371675 Chr.4: 44448239-44448239 on Build GRCh38
rs764253928 Chr.4: 44448272-44448272 on Build GRCh38
rs141407421 Chr.4: 44448139-44448139 on Build GRCh38
rs753115913 Chr.4: 44448174-44448174 on Build GRCh38
rs569885993 Chr.4: 44448287-44448287 on Build GRCh38
rs1016600855 Chr.4: 44448193-44448193 on Build GRCh38
rs964170538 Chr.4: 44448199-44448199 on Build GRCh38
rs372659933 Chr.4: 44448222-44448222 on Build GRCh38
rs780781463 Chr.4: 44448169-44448169 on Build GRCh38
rs754671545 Chr.4: 44448208-44448208 on Build GRCh38
rs370011280 Chr.4: 44448173-44448173 on Build GRCh38
rs138052184 Chr.4: 44448162-44448162 on Build GRCh38
rs766947923 Chr.4: 44448104-44448104 on Build GRCh38
rs773798880 Chr.4: 44448081-44448081 on Build GRCh38
rs761736071 Chr.4: 44448276-44448276 on Build GRCh38
rs377066294 Chr.4: 44448226-44448226 on Build GRCh38
rs77455244 Chr.4: 44448198-44448198 on Build GRCh38
rs760280100 Chr.4: 44448165-44448165 on Build GRCh38
rs763360875 Chr.4: 44448270-44448270 on Build GRCh38
rs763154094 Chr.4: 44448176-44448176 on Build GRCh38
rs138935237 Chr.4: 44448114-44448114 on Build GRCh38
rs1047487576 Chr.4: 44448167-44448167 on Build GRCh38
rs149503892 Chr.4: 44448164-44448164 on Build GRCh38
rs761252090 Chr.4: 44448100-44448100 on Build GRCh38
rs917687346 Chr.4: 44448123-44448123 on Build GRCh38
rs200405668 Chr.4: 44448166-44448166 on Build GRCh38
rs949124406 Chr.4: 44448130-44448130 on Build GRCh38
rs144922333 Chr.4: 44448149-44448149 on Build GRCh38
rs146302810 Chr.4: 44448105-44448105 on Build GRCh38
rs770349626 Chr.4: 44448266-44448266 on Build GRCh38
rs754941222 Chr.4: 44448285-44448285 on Build GRCh38

View Full Product Details