Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10287912-10288167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM4
Entrez Gene ID
3386
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
Gene Aliases
CD242, LW
UniGene
Hs.706750
Gene Chromosome Location
Chr.19: 10286967-10288584 on Build GRCh38
Transcripts
NM_001039132.2 NM_001544.4 NM_022377.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4073781 c.812C>T p.A271V Chr.19: 10288100-10288100 on Build GRCh38
5016610 c.787_788insA p.S264fs*>10 Chr.19: 10288152-10288153 on Build GRCh38
4804167 c.662G>T p.R221L Chr.19: 10288027-10288027 on Build GRCh38
5474039 c.807C>A p.S269S Chr.19: 10288095-10288095 on Build GRCh38
5933821 c.702G>A p.W234* Chr.19: 10287990-10287990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919910191 Chr.19: 10287939-10287939 on Build GRCh38
rs775968659 Chr.19: 10288059-10288059 on Build GRCh38
rs375791254 Chr.19: 10288121-10288121 on Build GRCh38
rs75264555 Chr.19: 10288072-10288072 on Build GRCh38
rs866455292 Chr.19: 10288089-10288089 on Build GRCh38
rs781281480 Chr.19: 10288131-10288131 on Build GRCh38
rs949742878 Chr.19: 10288074-10288074 on Build GRCh38
rs762687841 Chr.19: 10287929-10287929 on Build GRCh38
rs747607311 Chr.19: 10288134-10288134 on Build GRCh38
rs369632584 Chr.19: 10287976-10287976 on Build GRCh38
rs907128907 Chr.19: 10287968-10287968 on Build GRCh38
rs759826978 Chr.19: 10287959-10287959 on Build GRCh38
rs758114506 Chr.19: 10287999-10287999 on Build GRCh38
rs188777743 Chr.19: 10287973-10287973 on Build GRCh38
rs1021353307 Chr.19: 10288014-10288014 on Build GRCh38
rs200228336 Chr.19: 10287981-10287981 on Build GRCh38
rs924394713 Chr.19: 10288127-10288127 on Build GRCh38
rs201399464 Chr.19: 10288138-10288138 on Build GRCh38
rs780802328 Chr.19: 10288020-10288020 on Build GRCh38
rs754832864 Chr.19: 10288112-10288112 on Build GRCh38
rs113892276 Chr.19: 10288137-10288137 on Build GRCh38
rs779966296 Chr.19: 10288003-10288003 on Build GRCh38
rs373582901 Chr.19: 10288002-10288002 on Build GRCh38
rs930003017 Chr.19: 10287940-10287940 on Build GRCh38
rs145209145 Chr.19: 10288095-10288095 on Build GRCh38
rs753933674 Chr.19: 10287985-10287985 on Build GRCh38
rs773015279 Chr.19: 10288032-10288032 on Build GRCh38
rs768014944 Chr.19: 10287971-10287971 on Build GRCh38
rs915787425 Chr.19: 10288068-10288068 on Build GRCh38
rs750159706 Chr.19: 10287997-10287997 on Build GRCh38
rs370615268 Chr.19: 10288065-10288065 on Build GRCh38
rs757428309 Chr.19: 10287992-10287992 on Build GRCh38
rs372747645 Chr.19: 10287958-10287958 on Build GRCh38
rs774202084 Chr.19: 10288060-10288060 on Build GRCh38
rs768689512 Chr.19: 10288019-10288019 on Build GRCh38
rs184847594 Chr.19: 10288098-10288098 on Build GRCh38
rs761536113 Chr.19: 10287957-10287957 on Build GRCh38
rs764133200 Chr.19: 10287980-10287980 on Build GRCh38
rs377076712 Chr.19: 10288111-10288111 on Build GRCh38
rs772322610 Chr.19: 10288050-10288050 on Build GRCh38
rs5030385 Chr.19: 10287932-10287932 on Build GRCh38
rs779110405 Chr.19: 10287996-10287996 on Build GRCh38
rs762728283 Chr.19: 10288036-10288036 on Build GRCh38
rs753845734 Chr.19: 10288073-10288073 on Build GRCh38
rs370045160 Chr.19: 10287934-10287934 on Build GRCh38
rs867115356 Chr.19: 10288022-10288022 on Build GRCh38
rs761892488 Chr.19: 10288079-10288079 on Build GRCh38
rs747694527 Chr.19: 10288030-10288030 on Build GRCh38
rs115855638 Chr.19: 10287987-10287987 on Build GRCh38
rs140289465 Chr.19: 10288031-10288031 on Build GRCh38
rs758686500 Chr.19: 10288108-10288108 on Build GRCh38
rs955622882 Chr.19: 10288124-10288124 on Build GRCh38
rs761034821 Chr.19: 10288063-10288063 on Build GRCh38
rs539994937 Chr.19: 10288005-10288005 on Build GRCh38

View Full Product Details