Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134807496-134807767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122C
Entrez Gene ID
159091
Gene Name
family with sequence similarity 122C
Gene Aliases
-
UniGene
Hs.269127
Gene Chromosome Location
Chr.X: 134796413-134855137 on Build GRCh38
Transcripts
NM_001170779.1 NM_001170780.1 NM_001170781.1 NM_001170782.1 NM_001170783.1 NM_001170784.1 NM_138819.3 XM_005262382.1 XM_005262383.2 XM_005262384.4 XM_005262386.3 XM_005262387.3 XM_006724734.3 XM_006724735.1 XM_006724736.2 XM_011531295.1 XM_011531296.2 XM_011531297.2 XM_011531298.2 XM_011531305.2 XM_011531306.2 XM_017029308.1 XM_017029310.1 XM_017029311.1

SNP Detail

SNP ID Chromosome Location
rs773028001 Chr.X: 134807677-134807677 on Build GRCh38
rs746127027 Chr.X: 134807633-134807633 on Build GRCh38
rs865946484 Chr.X: 134807570-134807570 on Build GRCh38
rs370722079 Chr.X: 134807592-134807592 on Build GRCh38
rs201995773 Chr.X: 134807594-134807594 on Build GRCh38
rs775830736 Chr.X: 134807652-134807652 on Build GRCh38
rs755090654 Chr.X: 134807566-134807566 on Build GRCh38
rs770348329 Chr.X: 134807678-134807680 on Build GRCh38
rs764756069 Chr.X: 134807580-134807580 on Build GRCh38
rs757580800 Chr.X: 134807615-134807615 on Build GRCh38
rs764928413 Chr.X: 134807707-134807707 on Build GRCh38
rs943035228 Chr.X: 134807729-134807729 on Build GRCh38
rs370315393 Chr.X: 134807716-134807716 on Build GRCh38
rs760450594 Chr.X: 134807528-134807528 on Build GRCh38
rs752255352 Chr.X: 134807582-134807582 on Build GRCh38
rs768773441 Chr.X: 134807671-134807671 on Build GRCh38
rs766131444 Chr.X: 134807546-134807546 on Build GRCh38
rs377696356 Chr.X: 134807704-134807704 on Build GRCh38
rs983062191 Chr.X: 134807640-134807640 on Build GRCh38
rs913764456 Chr.X: 134807681-134807681 on Build GRCh38
rs773184057 Chr.X: 134807526-134807526 on Build GRCh38
rs760602908 Chr.X: 134807685-134807685 on Build GRCh38
rs749433780 Chr.X: 134807653-134807653 on Build GRCh38
rs924934037 Chr.X: 134807726-134807726 on Build GRCh38
rs750863628 Chr.X: 134807747-134807747 on Build GRCh38
rs746427517 Chr.X: 134807674-134807675 on Build GRCh38
rs781523284 Chr.X: 134807745-134807745 on Build GRCh38
rs201369085 Chr.X: 134807614-134807614 on Build GRCh38
rs757745326 Chr.X: 134807744-134807744 on Build GRCh38
rs374990973 Chr.X: 134807722-134807722 on Build GRCh38
rs776368516 Chr.X: 134807700-134807700 on Build GRCh38
rs143181255 Chr.X: 134807736-134807736 on Build GRCh38
rs150653042 Chr.X: 134807699-134807699 on Build GRCh38
rs781575411 Chr.X: 134807618-134807618 on Build GRCh38
rs754574795 Chr.X: 134807571-134807571 on Build GRCh38
rs770183324 Chr.X: 134807634-134807634 on Build GRCh38
rs763606643 Chr.X: 134807742-134807742 on Build GRCh38

View Full Product Details