Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129900325-129900598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP1
Entrez Gene ID
23048
Gene Name
formin binding protein 1
Gene Aliases
FBP17
UniGene
Hs.189409
Gene Chromosome Location
Chr.9: 129887187-130053846 on Build GRCh38
Transcripts
NM_015033.2 XM_005251815.2 XM_005251820.2 XM_005251821.2 XM_005251822.2 XM_005251823.2 XM_005251824.2 XM_005251825.2 XM_005251826.2 XM_005251827.2 XM_005251828.2 XM_005251829.2 XM_005251830.2 XM_005251831.2 XM_005251832.2 XM_005251833.2 XM_005251834.2 XM_006717016.2 XM_011518399.2 XM_011518400.2 XM_011518401.1 XM_011518402.1 XM_017014487.1 XM_017014488.1 XM_017014489.1 XM_017014490.1 XM_017014491.1 XM_017014492.1 XM_017014493.1 XM_017014494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5482247 c.1543C>T p.R515C Chr.9: 129900433-129900433 on Build GRCh38
6422966 c.1454G>A p.R485H Chr.9: 129900495-129900495 on Build GRCh38
5572695 c.1473C>T p.Y491Y Chr.9: 129900476-129900476 on Build GRCh38
455470 c.1469G>A p.S490N Chr.9: 129900507-129900507 on Build GRCh38
6422968 c.365G>A p.R122H Chr.9: 129900495-129900495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868352818 Chr.9: 129900392-129900392 on Build GRCh38
rs983038079 Chr.9: 129900569-129900569 on Build GRCh38
rs377021873 Chr.9: 129900565-129900565 on Build GRCh38
rs745482274 Chr.9: 129900411-129900411 on Build GRCh38
rs759741577 Chr.9: 129900517-129900517 on Build GRCh38
rs748125218 Chr.9: 129900578-129900578 on Build GRCh38
rs778380891 Chr.9: 129900570-129900570 on Build GRCh38
rs41279168 Chr.9: 129900443-129900443 on Build GRCh38
rs371932612 Chr.9: 129900498-129900498 on Build GRCh38
rs753995539 Chr.9: 129900510-129900510 on Build GRCh38
rs541597740 Chr.9: 129900505-129900505 on Build GRCh38
rs780950627 Chr.9: 129900408-129900408 on Build GRCh38
rs1023000 Chr.9: 129900507-129900507 on Build GRCh38
rs750723554 Chr.9: 129900454-129900454 on Build GRCh38
rs774310830 Chr.9: 129900496-129900496 on Build GRCh38
rs766666106 Chr.9: 129900509-129900509 on Build GRCh38
rs374564198 Chr.9: 129900486-129900486 on Build GRCh38
rs892285187 Chr.9: 129900506-129900506 on Build GRCh38
rs769413203 Chr.9: 129900417-129900417 on Build GRCh38
rs199695866 Chr.9: 129900407-129900407 on Build GRCh38
rs989440986 Chr.9: 129900354-129900354 on Build GRCh38
rs111319812 Chr.9: 129900546-129900546 on Build GRCh38
rs769274525 Chr.9: 129900491-129900491 on Build GRCh38
rs1050381060 Chr.9: 129900430-129900430 on Build GRCh38
rs774968395 Chr.9: 129900419-129900419 on Build GRCh38
rs955855132 Chr.9: 129900567-129900567 on Build GRCh38
rs548685688 Chr.9: 129900539-129900539 on Build GRCh38
rs367642326 Chr.9: 129900494-129900494 on Build GRCh38
rs779545854 Chr.9: 129900493-129900493 on Build GRCh38
rs200153012 Chr.9: 129900442-129900442 on Build GRCh38
rs574719725 Chr.9: 129900523-129900523 on Build GRCh38
rs908708546 Chr.9: 129900360-129900360 on Build GRCh38
rs1020941400 Chr.9: 129900577-129900577 on Build GRCh38
rs779421843 Chr.9: 129900403-129900403 on Build GRCh38
rs753900226 Chr.9: 129900471-129900471 on Build GRCh38
rs758841600 Chr.9: 129900476-129900476 on Build GRCh38
rs751833858 Chr.9: 129900426-129900426 on Build GRCh38
rs765380408 Chr.9: 129900522-129900522 on Build GRCh38
rs755482250 Chr.9: 129900387-129900387 on Build GRCh38
rs755571290 Chr.9: 129900472-129900472 on Build GRCh38
rs768130616 Chr.9: 129900495-129900495 on Build GRCh38
rs762892081 Chr.9: 129900420-129900420 on Build GRCh38
rs778000510 Chr.9: 129900484-129900484 on Build GRCh38
rs552645500 Chr.9: 129900475-129900475 on Build GRCh38
rs764532868 Chr.9: 129900529-129900529 on Build GRCh38
rs767185137 Chr.9: 129900444-129900444 on Build GRCh38
rs748906431 Chr.9: 129900564-129900564 on Build GRCh38
rs201780050 Chr.9: 129900497-129900497 on Build GRCh38
rs760790973 Chr.9: 129900455-129900455 on Build GRCh38
rs367878072 Chr.9: 129900511-129900511 on Build GRCh38

View Full Product Details