Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43972214-43972483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH2
Entrez Gene ID
1802
Gene Name
DPH2 homolog
Gene Aliases
DPH2L2
UniGene
Hs.632398
Gene Chromosome Location
Chr.1: 43969979-43973371 on Build GRCh38
Transcripts
NM_001039589.1 NM_001319165.1 NM_001319166.1 NM_001319167.1 NM_001319168.1 NM_001319169.1 NM_001319170.1 NM_001319171.1 NM_001384.4 XM_017000502.1 XM_017000503.1 XM_017000504.1 XM_017000505.1 XM_017000506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6191806 c.1414G>A p.A472T Chr.1: 43972483-43972483 on Build GRCh38
4679242 c.1228G>A p.V410M Chr.1: 43972217-43972217 on Build GRCh38
6226162 c.1258C>T p.P420S Chr.1: 43972247-43972247 on Build GRCh38
681243 c.1227C>T p.D409D Chr.1: 43972216-43972216 on Build GRCh38
358111 c.1363C>A p.R455R Chr.1: 43972432-43972432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368133869 Chr.1: 43972358-43972358 on Build GRCh38
rs776786597 Chr.1: 43972256-43972256 on Build GRCh38
rs372328715 Chr.1: 43972432-43972432 on Build GRCh38
rs759382348 Chr.1: 43972387-43972387 on Build GRCh38
rs148843738 Chr.1: 43972453-43972453 on Build GRCh38
rs758874474 Chr.1: 43972400-43972400 on Build GRCh38
rs758850602 Chr.1: 43972344-43972344 on Build GRCh38
rs766229067 Chr.1: 43972386-43972386 on Build GRCh38
rs755795415 Chr.1: 43972396-43972396 on Build GRCh38
rs766145822 Chr.1: 43972298-43972298 on Build GRCh38
rs747367994 Chr.1: 43972406-43972406 on Build GRCh38
rs759294446 Chr.1: 43972437-43972437 on Build GRCh38
rs754556372 Chr.1: 43972300-43972300 on Build GRCh38
rs770120491 Chr.1: 43972463-43972463 on Build GRCh38
rs750428481 Chr.1: 43972273-43972273 on Build GRCh38
rs571042588 Chr.1: 43972346-43972346 on Build GRCh38
rs149715554 Chr.1: 43972433-43972433 on Build GRCh38
rs773272790 Chr.1: 43972423-43972423 on Build GRCh38
rs763610967 Chr.1: 43972398-43972398 on Build GRCh38
rs762053576 Chr.1: 43972258-43972258 on Build GRCh38
rs762628746 Chr.1: 43972287-43972287 on Build GRCh38
rs765074908 Chr.1: 43972260-43972260 on Build GRCh38
rs772526649 Chr.1: 43972240-43972240 on Build GRCh38
rs781538048 Chr.1: 43972418-43972418 on Build GRCh38
rs375395515 Chr.1: 43972462-43972462 on Build GRCh38
rs775285736 Chr.1: 43972451-43972451 on Build GRCh38
rs776525529 Chr.1: 43972375-43972375 on Build GRCh38
rs752386249 Chr.1: 43972394-43972394 on Build GRCh38
rs777052949 Chr.1: 43972378-43972378 on Build GRCh38
rs780494389 Chr.1: 43972402-43972402 on Build GRCh38
rs753387029 Chr.1: 43972456-43972456 on Build GRCh38
rs72890649 Chr.1: 43972384-43972384 on Build GRCh38
rs533385756 Chr.1: 43972382-43972382 on Build GRCh38
rs35286676 Chr.1: 43972415-43972415 on Build GRCh38
rs755668773 Chr.1: 43972325-43972325 on Build GRCh38
rs199586736 Chr.1: 43972320-43972320 on Build GRCh38
rs763659580 Chr.1: 43972454-43972454 on Build GRCh38
rs775750438 Chr.1: 43972241-43972241 on Build GRCh38
rs368226729 Chr.1: 43972393-43972393 on Build GRCh38
rs760930824 Chr.1: 43972242-43972242 on Build GRCh38
rs751321281 Chr.1: 43972299-43972299 on Build GRCh38
rs748345159 Chr.1: 43972420-43972420 on Build GRCh38
rs761290368 Chr.1: 43972457-43972457 on Build GRCh38
rs767220738 Chr.1: 43972301-43972301 on Build GRCh38
rs373700666 Chr.1: 43972281-43972281 on Build GRCh38
rs767259308 Chr.1: 43972444-43972444 on Build GRCh38
rs779256768 Chr.1: 43972340-43972340 on Build GRCh38
rs200212326 Chr.1: 43972385-43972385 on Build GRCh38
rs769917567 Chr.1: 43972422-43972422 on Build GRCh38
rs769015609 Chr.1: 43972247-43972247 on Build GRCh38

View Full Product Details