Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77243655-77243928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD4
Entrez Gene ID
220032
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases
GDE6
UniGene
Hs.249795
Gene Chromosome Location
Chr.11: 77216558-77301699 on Build GRCh38
Transcripts
NM_182833.1 XM_011544826.1 XM_011544833.1 XM_011544834.1 XM_017017348.1 XM_017017349.1 XM_017017350.1 XM_017017351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4424729 c.1171G>A p.E391K Chr.11: 77243764-77243764 on Build GRCh38
7229161 c.1168A>G p.I390V Chr.11: 77243767-77243767 on Build GRCh38
7229328 c.1147C>T p.H383Y Chr.11: 77243788-77243788 on Build GRCh38
5531657 c.1100C>T p.P367L Chr.11: 77243835-77243835 on Build GRCh38
3935638 c.1165T>A p.S389T Chr.11: 77243770-77243770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781532010 Chr.11: 77243707-77243707 on Build GRCh38
rs144726122 Chr.11: 77243693-77243693 on Build GRCh38
rs758407571 Chr.11: 77243690-77243690 on Build GRCh38
rs139949435 Chr.11: 77243816-77243816 on Build GRCh38
rs761293382 Chr.11: 77243737-77243737 on Build GRCh38
rs769582672 Chr.11: 77243721-77243721 on Build GRCh38
rs748008884 Chr.11: 77243875-77243875 on Build GRCh38
rs770655172 Chr.11: 77243893-77243893 on Build GRCh38
rs750660414 Chr.11: 77243774-77243774 on Build GRCh38
rs764691124 Chr.11: 77243828-77243828 on Build GRCh38
rs376504485 Chr.11: 77243759-77243759 on Build GRCh38
rs750526391 Chr.11: 77243848-77243848 on Build GRCh38
rs763195741 Chr.11: 77243827-77243827 on Build GRCh38
rs778847034 Chr.11: 77243874-77243874 on Build GRCh38
rs1029143976 Chr.11: 77243795-77243795 on Build GRCh38
rs530921395 Chr.11: 77243864-77243864 on Build GRCh38
rs947886229 Chr.11: 77243742-77243742 on Build GRCh38
rs199757792 Chr.11: 77243807-77243807 on Build GRCh38
rs775969034 Chr.11: 77243825-77243825 on Build GRCh38
rs869236617 Chr.11: 77243744-77243744 on Build GRCh38
rs746583946 Chr.11: 77243906-77243906 on Build GRCh38
rs538219375 Chr.11: 77243689-77243693 on Build GRCh38
rs759903651 Chr.11: 77243753-77243753 on Build GRCh38
rs756018735 Chr.11: 77243848-77243849 on Build GRCh38
rs749182440 Chr.11: 77243729-77243729 on Build GRCh38
rs936015195 Chr.11: 77243786-77243786 on Build GRCh38
rs11237145 Chr.11: 77243767-77243767 on Build GRCh38
rs200553823 Chr.11: 77243879-77243879 on Build GRCh38
rs767202060 Chr.11: 77243740-77243740 on Build GRCh38
rs754693771 Chr.11: 77243872-77243872 on Build GRCh38
rs371415425 Chr.11: 77243899-77243899 on Build GRCh38
rs754733637 Chr.11: 77243730-77243730 on Build GRCh38
rs765567778 Chr.11: 77243754-77243754 on Build GRCh38
rs142214451 Chr.11: 77243819-77243819 on Build GRCh38
rs34784710 Chr.11: 77243743-77243744 on Build GRCh38
rs199737312 Chr.11: 77243768-77243768 on Build GRCh38
rs1025555367 Chr.11: 77243685-77243685 on Build GRCh38
rs189759383 Chr.11: 77243822-77243822 on Build GRCh38
rs761997899 Chr.11: 77243834-77243834 on Build GRCh38
rs753710057 Chr.11: 77243871-77243871 on Build GRCh38
rs547010874 Chr.11: 77243905-77243905 on Build GRCh38
rs777973148 Chr.11: 77243691-77243691 on Build GRCh38
rs1045537477 Chr.11: 77243705-77243705 on Build GRCh38
rs201721341 Chr.11: 77243824-77243824 on Build GRCh38
rs186959963 Chr.11: 77243678-77243678 on Build GRCh38
rs766039759 Chr.11: 77243688-77243691 on Build GRCh38
rs202143817 Chr.11: 77243745-77243745 on Build GRCh38
rs752578399 Chr.11: 77243747-77243748 on Build GRCh38
rs75071001 Chr.11: 77243892-77243892 on Build GRCh38
rs373470613 Chr.11: 77243812-77243812 on Build GRCh38
rs536599606 Chr.11: 77243833-77243833 on Build GRCh38
rs11237146 Chr.11: 77243788-77243788 on Build GRCh38
rs774243373 Chr.11: 77243813-77243813 on Build GRCh38
rs777319542 Chr.11: 77243881-77243881 on Build GRCh38
rs917781248 Chr.11: 77243862-77243862 on Build GRCh38
rs762616901 Chr.11: 77243683-77243686 on Build GRCh38
rs779968297 Chr.11: 77243803-77243803 on Build GRCh38
rs757600977 Chr.11: 77243699-77243699 on Build GRCh38
rs779270640 Chr.11: 77243815-77243815 on Build GRCh38
rs7104577 Chr.11: 77243853-77243853 on Build GRCh38
rs76229203 Chr.11: 77243744-77243745 on Build GRCh38
rs139634188 Chr.11: 77243733-77243733 on Build GRCh38
rs745689001 Chr.11: 77243714-77243714 on Build GRCh38
rs757371607 Chr.11: 77243778-77243778 on Build GRCh38
rs905222052 Chr.11: 77243718-77243718 on Build GRCh38
rs781587093 Chr.11: 77243779-77243779 on Build GRCh38
rs775303267 Chr.11: 77243727-77243727 on Build GRCh38
rs763398093 Chr.11: 77243900-77243900 on Build GRCh38
rs746030370 Chr.11: 77243780-77243780 on Build GRCh38

View Full Product Details