Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48639622-48639849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR3
Entrez Gene ID
1951
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
Gene Aliases
ADGRC3, CDHF11, EGFL1, FMI1, HFMI1, MEGF2, RESDA1
UniGene
Hs.631926
Gene Chromosome Location
Chr.3: 48636463-48662915 on Build GRCh38
Transcripts
NM_001407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3070845 c.9866G>A p.R3289H Chr.3: 48639719-48639719 on Build GRCh38
6626337 c.9891G>A p.T3297T Chr.3: 48639709-48639709 on Build GRCh38
3070844 c.9875C>T p.T3292M Chr.3: 48639710-48639710 on Build GRCh38
6626336 c.9876G>A p.T3292T Chr.3: 48639709-48639709 on Build GRCh38
5927280 c.9755C>T p.S3252F Chr.3: 48639845-48639845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013283027 Chr.3: 48639676-48639676 on Build GRCh38
rs895399052 Chr.3: 48639651-48639651 on Build GRCh38
rs746226586 Chr.3: 48639713-48639713 on Build GRCh38
rs750689389 Chr.3: 48639824-48639824 on Build GRCh38
rs111652964 Chr.3: 48639748-48639748 on Build GRCh38
rs761603990 Chr.3: 48639656-48639656 on Build GRCh38
rs779197221 Chr.3: 48639745-48639745 on Build GRCh38
rs146846419 Chr.3: 48639746-48639746 on Build GRCh38
rs751231826 Chr.3: 48639669-48639669 on Build GRCh38
rs990335876 Chr.3: 48639733-48639733 on Build GRCh38
rs754733471 Chr.3: 48639673-48639673 on Build GRCh38
rs770923405 Chr.3: 48639799-48639799 on Build GRCh38
rs756876883 Chr.3: 48639664-48639664 on Build GRCh38
rs61729239 Chr.3: 48639685-48639685 on Build GRCh38
rs769607301 Chr.3: 48639787-48639787 on Build GRCh38
rs767220916 Chr.3: 48639725-48639725 on Build GRCh38
rs373106405 Chr.3: 48639703-48639703 on Build GRCh38
rs769503815 Chr.3: 48639712-48639712 on Build GRCh38
rs931538196 Chr.3: 48639806-48639806 on Build GRCh38
rs749243467 Chr.3: 48639798-48639798 on Build GRCh38
rs745459213 Chr.3: 48639772-48639772 on Build GRCh38
rs765631739 Chr.3: 48639823-48639823 on Build GRCh38
rs933359385 Chr.3: 48639811-48639811 on Build GRCh38
rs200610415 Chr.3: 48639810-48639810 on Build GRCh38
rs146685584 Chr.3: 48639720-48639720 on Build GRCh38
rs747037663 Chr.3: 48639761-48639761 on Build GRCh38
rs746402820 Chr.3: 48639695-48639695 on Build GRCh38
rs768801767 Chr.3: 48639706-48639706 on Build GRCh38
rs765842926 Chr.3: 48639667-48639667 on Build GRCh38
rs758898272 Chr.3: 48639754-48639754 on Build GRCh38
rs61729240 Chr.3: 48639681-48639681 on Build GRCh38
rs61729242 Chr.3: 48639758-48639758 on Build GRCh38
rs762482941 Chr.3: 48639666-48639666 on Build GRCh38
rs758629916 Chr.3: 48639659-48639659 on Build GRCh38
rs200592775 Chr.3: 48639719-48639719 on Build GRCh38
rs760303916 Chr.3: 48639821-48639821 on Build GRCh38
rs759067015 Chr.3: 48639813-48639813 on Build GRCh38
rs748354915 Chr.3: 48639785-48639785 on Build GRCh38
rs919997278 Chr.3: 48639803-48639803 on Build GRCh38
rs369105633 Chr.3: 48639686-48639686 on Build GRCh38
rs765167277 Chr.3: 48639662-48639662 on Build GRCh38
rs772935901 Chr.3: 48639792-48639792 on Build GRCh38
rs376186501 Chr.3: 48639710-48639710 on Build GRCh38
rs779436535 Chr.3: 48639694-48639694 on Build GRCh38
rs747229950 Chr.3: 48639705-48639705 on Build GRCh38
rs201850388 Chr.3: 48639724-48639724 on Build GRCh38
rs752429764 Chr.3: 48639729-48639729 on Build GRCh38
rs572407704 Chr.3: 48639739-48639739 on Build GRCh38
rs946244638 Chr.3: 48639763-48639763 on Build GRCh38
rs114411876 Chr.3: 48639709-48639709 on Build GRCh38
rs762844075 Chr.3: 48639717-48639717 on Build GRCh38
rs370984354 Chr.3: 48639818-48639818 on Build GRCh38
rs576543149 Chr.3: 48639817-48639817 on Build GRCh38
rs781349045 Chr.3: 48639781-48639781 on Build GRCh38
rs972850209 Chr.3: 48639786-48639786 on Build GRCh38
rs755076639 Chr.3: 48639767-48639767 on Build GRCh38
rs1042268919 Chr.3: 48639780-48639780 on Build GRCh38
rs377726340 Chr.3: 48639770-48639770 on Build GRCh38
rs1050853900 Chr.3: 48639657-48639657 on Build GRCh38
rs772171612 Chr.3: 48639702-48639702 on Build GRCh38

View Full Product Details