Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19062985-19063246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
UniGene
Hs.533468
Gene Chromosome Location
Chr.9: 19053137-19102942 on Build GRCh38
Transcripts
NM_001270890.1 NM_017645.4 XM_011517935.2 XM_017014830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71021 c.1627G>A p.E543K Chr.9: 19063010-19063010 on Build GRCh38
1461651 c.1588delA p.S530fs*12 Chr.9: 19063049-19063049 on Build GRCh38
752908 c.1529C>G p.S510* Chr.9: 19063108-19063108 on Build GRCh38
6115190 c.1605A>G p.K535K Chr.9: 19063032-19063032 on Build GRCh38
5876037 c.1594C>T p.P532S Chr.9: 19063043-19063043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755140796 Chr.9: 19063043-19063043 on Build GRCh38
rs147787866 Chr.9: 19063097-19063097 on Build GRCh38
rs917252880 Chr.9: 19063079-19063079 on Build GRCh38
rs778095707 Chr.9: 19063210-19063210 on Build GRCh38
rs773829089 Chr.9: 19063085-19063085 on Build GRCh38
rs770680634 Chr.9: 19063099-19063099 on Build GRCh38
rs199958062 Chr.9: 19063083-19063083 on Build GRCh38
rs146554163 Chr.9: 19063090-19063090 on Build GRCh38
rs34304297 Chr.9: 19063036-19063036 on Build GRCh38
rs868433253 Chr.9: 19063169-19063169 on Build GRCh38
rs751153471 Chr.9: 19063154-19063154 on Build GRCh38
rs774995466 Chr.9: 19063121-19063121 on Build GRCh38
rs771958444 Chr.9: 19063139-19063140 on Build GRCh38
rs199865674 Chr.9: 19063100-19063100 on Build GRCh38
rs756545345 Chr.9: 19063161-19063161 on Build GRCh38
rs771642835 Chr.9: 19063053-19063053 on Build GRCh38
rs13302748 Chr.9: 19063110-19063110 on Build GRCh38
rs868575536 Chr.9: 19063176-19063176 on Build GRCh38
rs762363345 Chr.9: 19063128-19063128 on Build GRCh38
rs755486457 Chr.9: 19063170-19063170 on Build GRCh38
rs996106216 Chr.9: 19063047-19063047 on Build GRCh38
rs766814464 Chr.9: 19063163-19063163 on Build GRCh38
rs764737381 Chr.9: 19063117-19063117 on Build GRCh38
rs780759471 Chr.9: 19063021-19063021 on Build GRCh38
rs779140647 Chr.9: 19063173-19063173 on Build GRCh38
rs375714104 Chr.9: 19063183-19063183 on Build GRCh38
rs758384751 Chr.9: 19063202-19063202 on Build GRCh38
rs960456873 Chr.9: 19063198-19063198 on Build GRCh38
rs756425032 Chr.9: 19063016-19063016 on Build GRCh38
rs763867424 Chr.9: 19063144-19063144 on Build GRCh38
rs746401959 Chr.9: 19063022-19063022 on Build GRCh38
rs34558496 Chr.9: 19063055-19063055 on Build GRCh38
rs532206494 Chr.9: 19063168-19063168 on Build GRCh38

View Full Product Details