Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32613856-32614113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC73
Entrez Gene ID
493860
Gene Name
coiled-coil domain containing 73
Gene Aliases
NY-SAR-79
UniGene
Hs.706808
Gene Chromosome Location
Chr.11: 32602080-32794658 on Build GRCh38
Transcripts
NM_001008391.3 XM_011520139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1684251 c.2235delA p.T747fs*6 Chr.11: 32614083-32614083 on Build GRCh38
3727147 c.2443G>A p.E815K Chr.11: 32613875-32613875 on Build GRCh38
3415808 c.2371G>T p.D791Y Chr.11: 32613947-32613947 on Build GRCh38
224715 c.2234G>A p.G745E Chr.11: 32614084-32614084 on Build GRCh38
926383 c.2413G>T p.E805* Chr.11: 32613905-32613905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779367830 Chr.11: 32614018-32614018 on Build GRCh38
rs202108382 Chr.11: 32614077-32614077 on Build GRCh38
rs760713150 Chr.11: 32613911-32613911 on Build GRCh38
rs537297161 Chr.11: 32614015-32614015 on Build GRCh38
rs201561301 Chr.11: 32614041-32614041 on Build GRCh38
rs762315078 Chr.11: 32614042-32614042 on Build GRCh38
rs369352957 Chr.11: 32613891-32613891 on Build GRCh38
rs752298919 Chr.11: 32613991-32613991 on Build GRCh38
rs1038893313 Chr.11: 32613933-32613933 on Build GRCh38
rs772319997 Chr.11: 32614033-32614033 on Build GRCh38
rs372961759 Chr.11: 32613895-32613895 on Build GRCh38
rs752386781 Chr.11: 32614083-32614083 on Build GRCh38
rs749779493 Chr.11: 32614073-32614073 on Build GRCh38
rs933291186 Chr.11: 32614011-32614011 on Build GRCh38
rs746170374 Chr.11: 32614030-32614030 on Build GRCh38
rs267602855 Chr.11: 32614062-32614062 on Build GRCh38
rs762738980 Chr.11: 32613945-32613945 on Build GRCh38
rs568361428 Chr.11: 32613913-32613913 on Build GRCh38
rs767349774 Chr.11: 32614079-32614079 on Build GRCh38
rs751268267 Chr.11: 32613960-32613960 on Build GRCh38
rs778484527 Chr.11: 32614001-32614001 on Build GRCh38
rs746107663 Chr.11: 32613889-32613889 on Build GRCh38
rs754384702 Chr.11: 32614010-32614010 on Build GRCh38
rs766197622 Chr.11: 32613953-32613953 on Build GRCh38
rs774247652 Chr.11: 32614078-32614078 on Build GRCh38
rs759280522 Chr.11: 32613969-32613969 on Build GRCh38
rs773185905 Chr.11: 32613932-32613932 on Build GRCh38
rs756566586 Chr.11: 32613884-32613888 on Build GRCh38
rs768734209 Chr.11: 32614054-32614054 on Build GRCh38
rs755684589 Chr.11: 32613995-32613995 on Build GRCh38
rs35425789 Chr.11: 32614083-32614084 on Build GRCh38
rs762976217 Chr.11: 32614076-32614076 on Build GRCh38
rs866552882 Chr.11: 32613924-32613924 on Build GRCh38
rs747148105 Chr.11: 32614047-32614047 on Build GRCh38
rs200376580 Chr.11: 32614063-32614063 on Build GRCh38
rs767260028 Chr.11: 32613977-32613977 on Build GRCh38
rs780662324 Chr.11: 32613903-32613908 on Build GRCh38

View Full Product Details