Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78580748-78581009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3504312 c.175G>A p.E59K Chr.15: 78580879-78580879 on Build GRCh38
6246175 c.188G>T p.R63L Chr.15: 78580892-78580892 on Build GRCh38
4057149 c.258+2T>C p.? Chr.15: 78580964-78580964 on Build GRCh38
3504314 c.192T>A p.P64P Chr.15: 78580896-78580896 on Build GRCh38
7282137 c.166C>G p.Q56E Chr.15: 78580870-78580870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745545261 Chr.15: 78580867-78580867 on Build GRCh38
rs755693811 Chr.15: 78580969-78580969 on Build GRCh38
rs772549498 Chr.15: 78580823-78580823 on Build GRCh38
rs747798581 Chr.15: 78580959-78580967 on Build GRCh38
rs183719313 Chr.15: 78580891-78580891 on Build GRCh38
rs760981472 Chr.15: 78580905-78580905 on Build GRCh38
rs199587175 Chr.15: 78580942-78580942 on Build GRCh38
rs533238214 Chr.15: 78580951-78580951 on Build GRCh38
rs773093762 Chr.15: 78580954-78580954 on Build GRCh38
rs771858151 Chr.15: 78580953-78580953 on Build GRCh38
rs750725297 Chr.15: 78580797-78580797 on Build GRCh38
rs767203807 Chr.15: 78580792-78580792 on Build GRCh38
rs748585735 Chr.15: 78580816-78580816 on Build GRCh38
rs201999654 Chr.15: 78580822-78580822 on Build GRCh38
rs201569403 Chr.15: 78580940-78580940 on Build GRCh38
rs56351164 Chr.15: 78580930-78580930 on Build GRCh38
rs376466023 Chr.15: 78580808-78580808 on Build GRCh38
rs79835149 Chr.15: 78580878-78580878 on Build GRCh38
rs760430286 Chr.15: 78580960-78580960 on Build GRCh38
rs145146999 Chr.15: 78580920-78580920 on Build GRCh38
rs901383530 Chr.15: 78580968-78580968 on Build GRCh38
rs754506075 Chr.15: 78580856-78580856 on Build GRCh38
rs780642757 Chr.15: 78580789-78580789 on Build GRCh38
rs368213023 Chr.15: 78580829-78580829 on Build GRCh38
rs200186728 Chr.15: 78580899-78580899 on Build GRCh38
rs200324605 Chr.15: 78580935-78580935 on Build GRCh38
rs754032640 Chr.15: 78580907-78580907 on Build GRCh38
rs372781673 Chr.15: 78580903-78580903 on Build GRCh38
rs748935802 Chr.15: 78580944-78580944 on Build GRCh38
rs201040941 Chr.15: 78580871-78580871 on Build GRCh38
rs202110255 Chr.15: 78580874-78580874 on Build GRCh38
rs543210727 Chr.15: 78580879-78580879 on Build GRCh38
rs199919224 Chr.15: 78580850-78580850 on Build GRCh38
rs555895292 Chr.15: 78580977-78580983 on Build GRCh38
rs751094077 Chr.15: 78580823-78580825 on Build GRCh38
rs377184396 Chr.15: 78580914-78580914 on Build GRCh38
rs201556084 Chr.15: 78580904-78580904 on Build GRCh38
rs756226727 Chr.15: 78580798-78580798 on Build GRCh38
rs1050456017 Chr.15: 78580973-78580973 on Build GRCh38
rs768348027 Chr.15: 78580946-78580946 on Build GRCh38
rs200129149 Chr.15: 78580876-78580876 on Build GRCh38
rs367611089 Chr.15: 78580915-78580919 on Build GRCh38
rs542540729 Chr.15: 78580810-78580810 on Build GRCh38
rs202057419 Chr.15: 78580892-78580892 on Build GRCh38
rs755103444 Chr.15: 78580911-78580911 on Build GRCh38
rs773931755 Chr.15: 78580950-78580950 on Build GRCh38
rs751511725 Chr.15: 78580784-78580784 on Build GRCh38
rs553845006 Chr.15: 78580789-78580789 on Build GRCh38
rs749900040 Chr.15: 78580939-78580939 on Build GRCh38
rs766144208 Chr.15: 78580965-78580965 on Build GRCh38
rs778220199 Chr.15: 78580931-78580931 on Build GRCh38
rs200692387 Chr.15: 78580805-78580805 on Build GRCh38
rs569207 Chr.15: 78580777-78580777 on Build GRCh38
rs749320132 Chr.15: 78580806-78580806 on Build GRCh38
rs963252235 Chr.15: 78580938-78580938 on Build GRCh38

View Full Product Details