Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28551574-28551799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD3
Entrez Gene ID
7976
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3
UniGene
Hs.40735
Gene Chromosome Location
Chr.8: 28494205-28574268 on Build GRCh38
Transcripts
NM_017412.3 NM_145866.1 XM_017013841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406832 c.1421G>A p.R474H Chr.8: 28551619-28551619 on Build GRCh38
5880178 c.1481C>T p.S494F Chr.8: 28551679-28551679 on Build GRCh38
275237 c.1506G>T p.K502N Chr.8: 28551704-28551704 on Build GRCh38
6668151 c.1541G>A p.R514H Chr.8: 28551739-28551739 on Build GRCh38
454407 c.1409C>T p.T470I Chr.8: 28551607-28551607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760470003 Chr.8: 28551704-28551704 on Build GRCh38
rs766557184 Chr.8: 28551666-28551666 on Build GRCh38
rs74764832 Chr.8: 28551766-28551766 on Build GRCh38
rs757328457 Chr.8: 28551723-28551723 on Build GRCh38
rs562550070 Chr.8: 28551611-28551611 on Build GRCh38
rs139570173 Chr.8: 28551739-28551739 on Build GRCh38
rs769861018 Chr.8: 28551647-28551647 on Build GRCh38
rs781606805 Chr.8: 28551767-28551767 on Build GRCh38
rs768929181 Chr.8: 28551613-28551613 on Build GRCh38
rs930901306 Chr.8: 28551618-28551618 on Build GRCh38
rs773340232 Chr.8: 28551662-28551662 on Build GRCh38
rs201782719 Chr.8: 28551771-28551771 on Build GRCh38
rs755307590 Chr.8: 28551758-28551758 on Build GRCh38
rs373553202 Chr.8: 28551619-28551619 on Build GRCh38
rs944770329 Chr.8: 28551736-28551736 on Build GRCh38
rs920843630 Chr.8: 28551607-28551607 on Build GRCh38
rs764824535 Chr.8: 28551724-28551724 on Build GRCh38
rs771811674 Chr.8: 28551603-28551603 on Build GRCh38
rs531633095 Chr.8: 28551738-28551738 on Build GRCh38
rs764027388 Chr.8: 28551707-28551707 on Build GRCh38
rs376178388 Chr.8: 28551683-28551683 on Build GRCh38
rs762350717 Chr.8: 28551626-28551626 on Build GRCh38
rs762956166 Chr.8: 28551664-28551664 on Build GRCh38
rs753794190 Chr.8: 28551708-28551708 on Build GRCh38
rs779583947 Chr.8: 28551606-28551606 on Build GRCh38
rs749886914 Chr.8: 28551734-28551734 on Build GRCh38

View Full Product Details