Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9418533-9418774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB1BP1
Entrez Gene ID
9270
Gene Name
integrin subunit beta 1 binding protein 1
Gene Aliases
ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
UniGene
Hs.467662
Gene Chromosome Location
Chr.2: 9405681-9423569 on Build GRCh38
Transcripts
NM_001319066.1 NM_001319067.1 NM_001319068.1 NM_001319069.1 NM_001319070.1 NM_004763.4 NM_022334.4 NR_134951.1 NR_134952.1 NR_134953.1 NR_134954.1 NR_134955.1 NR_134956.1 NR_134957.1 XM_005246183.4 XM_005246184.4 XM_005246185.4 XM_005246189.4 XM_006711903.3 XM_011510416.2 XM_017005267.1 XM_017005268.1 XM_017005269.1 XM_017005270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6688174 c.1A>G p.M1V Chr.2: 9418697-9418697 on Build GRCh38
4096267 c.45A>T p.Q15H Chr.2: 9418653-9418653 on Build GRCh38
5611529 c.72+2_72+3TA>AG p.? Chr.2: 9418623-9418624 on Build GRCh38
180361 c.51C>T p.S17S Chr.2: 9418647-9418647 on Build GRCh38
1130721 c.14G>A p.G5D Chr.2: 9418684-9418684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769716068 Chr.2: 9418704-9418704 on Build GRCh38
rs972264938 Chr.2: 9418557-9418557 on Build GRCh38
rs933247565 Chr.2: 9418583-9418583 on Build GRCh38
rs765475880 Chr.2: 9418656-9418656 on Build GRCh38
rs963518635 Chr.2: 9418553-9418553 on Build GRCh38
rs143507814 Chr.2: 9418647-9418647 on Build GRCh38
rs200805544 Chr.2: 9418615-9418615 on Build GRCh38
rs372616266 Chr.2: 9418730-9418730 on Build GRCh38
rs747475755 Chr.2: 9418742-9418742 on Build GRCh38
rs1801980 Chr.2: 9418683-9418683 on Build GRCh38
rs781157155 Chr.2: 9418585-9418585 on Build GRCh38
rs745426916 Chr.2: 9418724-9418724 on Build GRCh38
rs748907150 Chr.2: 9418606-9418606 on Build GRCh38
rs576778262 Chr.2: 9418562-9418562 on Build GRCh38
rs923170234 Chr.2: 9418576-9418576 on Build GRCh38
rs369866159 Chr.2: 9418608-9418608 on Build GRCh38
rs757006502 Chr.2: 9418714-9418714 on Build GRCh38
rs763929319 Chr.2: 9418707-9418707 on Build GRCh38
rs200885957 Chr.2: 9418654-9418654 on Build GRCh38
rs749988877 Chr.2: 9418643-9418643 on Build GRCh38
rs755963107 Chr.2: 9418601-9418601 on Build GRCh38
rs200874587 Chr.2: 9418582-9418582 on Build GRCh38
rs773209764 Chr.2: 9418690-9418690 on Build GRCh38
rs267599492 Chr.2: 9418646-9418646 on Build GRCh38
rs752503922 Chr.2: 9418586-9418586 on Build GRCh38
rs747385235 Chr.2: 9418636-9418636 on Build GRCh38
rs778623007 Chr.2: 9418719-9418719 on Build GRCh38
rs374581919 Chr.2: 9418709-9418709 on Build GRCh38
rs777569864 Chr.2: 9418603-9418603 on Build GRCh38
rs772521477 Chr.2: 9418623-9418623 on Build GRCh38
rs776871592 Chr.2: 9418648-9418648 on Build GRCh38
rs756002029 Chr.2: 9418706-9418706 on Build GRCh38
rs776050652 Chr.2: 9418635-9418635 on Build GRCh38
rs757865713 Chr.2: 9418727-9418727 on Build GRCh38
rs1053877887 Chr.2: 9418676-9418676 on Build GRCh38

View Full Product Details