Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107218773-107219034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIH1D3
Entrez Gene ID
139212
Gene Name
PIH1 domain containing 3
Gene Aliases
CXorf41, NYSAR97
UniGene
Hs.128836
Gene Chromosome Location
Chr.X: 107206629-107244243 on Build GRCh38
Transcripts
NM_001169154.1 NM_173494.1 XM_011530855.2

SNP Detail

SNP ID Chromosome Location
rs762882859 Chr.X: 107218969-107218969 on Build GRCh38
rs747602928 Chr.X: 107218856-107218856 on Build GRCh38
rs147979953 Chr.X: 107218881-107218881 on Build GRCh38
rs373849863 Chr.X: 107218845-107218845 on Build GRCh38
rs749119819 Chr.X: 107218817-107218817 on Build GRCh38
rs778877277 Chr.X: 107218849-107218849 on Build GRCh38
rs746700360 Chr.X: 107218880-107218880 on Build GRCh38
rs760029187 Chr.X: 107218898-107218898 on Build GRCh38
rs765827471 Chr.X: 107218927-107218927 on Build GRCh38
rs764256526 Chr.X: 107218992-107218992 on Build GRCh38
rs779462284 Chr.X: 107218871-107218871 on Build GRCh38
rs377237630 Chr.X: 107218998-107218998 on Build GRCh38
rs777107680 Chr.X: 107218897-107218897 on Build GRCh38
rs768870165 Chr.X: 107218822-107218822 on Build GRCh38
rs772534884 Chr.X: 107218977-107218977 on Build GRCh38
rs80015507 Chr.X: 107218946-107218946 on Build GRCh38
rs771611594 Chr.X: 107218859-107218859 on Build GRCh38

View Full Product Details