Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36461336-36461575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN14
Entrez Gene ID
23562
Gene Name
claudin 14
Gene Aliases
DFNB29
UniGene
Hs.660278
Gene Chromosome Location
Chr.21: 36460621-36576569 on Build GRCh38
Transcripts
NM_001146077.1 NM_001146078.2 NM_001146079.1 NM_012130.3 NM_144492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681958 c.292G>A p.A98T Chr.21: 36461404-36461404 on Build GRCh38
4101483 c.250A>G p.M84V Chr.21: 36461446-36461446 on Build GRCh38
4167890 c.242G>A p.R81H Chr.21: 36461454-36461454 on Build GRCh38
6629965 c.336C>T p.P112P Chr.21: 36461360-36461360 on Build GRCh38
1307775 c.276C>T p.G92G Chr.21: 36461420-36461420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760769965 Chr.21: 36461501-36461501 on Build GRCh38
rs770829440 Chr.21: 36461443-36461443 on Build GRCh38
rs964200327 Chr.21: 36461424-36461424 on Build GRCh38
rs777960789 Chr.21: 36461501-36461501 on Build GRCh38
rs759313432 Chr.21: 36461404-36461404 on Build GRCh38
rs762368755 Chr.21: 36461409-36461409 on Build GRCh38
rs745601274 Chr.21: 36461527-36461527 on Build GRCh38
rs767565332 Chr.21: 36461487-36461487 on Build GRCh38
rs377019596 Chr.21: 36461374-36461374 on Build GRCh38
rs778842030 Chr.21: 36461525-36461525 on Build GRCh38
rs368027306 Chr.21: 36461454-36461454 on Build GRCh38
rs140257182 Chr.21: 36461380-36461380 on Build GRCh38
rs148223897 Chr.21: 36461511-36461511 on Build GRCh38
rs754350037 Chr.21: 36461360-36461360 on Build GRCh38
rs768019537 Chr.21: 36461537-36461537 on Build GRCh38
rs908752693 Chr.21: 36461475-36461475 on Build GRCh38
rs143628692 Chr.21: 36461455-36461455 on Build GRCh38
rs758724049 Chr.21: 36461416-36461416 on Build GRCh38
rs983012374 Chr.21: 36461464-36461464 on Build GRCh38
rs750946312 Chr.21: 36461411-36461411 on Build GRCh38
rs219779 Chr.21: 36461453-36461453 on Build GRCh38
rs557513586 Chr.21: 36461397-36461397 on Build GRCh38
rs138631461 Chr.21: 36461359-36461359 on Build GRCh38
rs773126341 Chr.21: 36461401-36461402 on Build GRCh38
rs577393948 Chr.21: 36461423-36461423 on Build GRCh38
rs573588226 Chr.21: 36461481-36461481 on Build GRCh38
rs746698259 Chr.21: 36461534-36461534 on Build GRCh38
rs74315437 Chr.21: 36461442-36461442 on Build GRCh38
rs767108790 Chr.21: 36461405-36461405 on Build GRCh38
rs113350364 Chr.21: 36461396-36461396 on Build GRCh38
rs756218341 Chr.21: 36461434-36461434 on Build GRCh38
rs371100799 Chr.21: 36461529-36461529 on Build GRCh38
rs755346413 Chr.21: 36461485-36461485 on Build GRCh38
rs775222236 Chr.21: 36461457-36461457 on Build GRCh38
rs751747842 Chr.21: 36461420-36461420 on Build GRCh38
rs765605373 Chr.21: 36461410-36461410 on Build GRCh38
rs760039736 Chr.21: 36461462-36461462 on Build GRCh38
rs780444223 Chr.21: 36461419-36461419 on Build GRCh38
rs374400022 Chr.21: 36461402-36461402 on Build GRCh38
rs764548516 Chr.21: 36461375-36461375 on Build GRCh38
rs555911451 Chr.21: 36461361-36461361 on Build GRCh38
rs981704813 Chr.21: 36461507-36461507 on Build GRCh38
rs752960573 Chr.21: 36461532-36461532 on Build GRCh38
rs781167430 Chr.21: 36461381-36461381 on Build GRCh38
rs947811327 Chr.21: 36461553-36461553 on Build GRCh38
rs777529810 Chr.21: 36461436-36461436 on Build GRCh38
rs763692550 Chr.21: 36461468-36461468 on Build GRCh38
rs865872458 Chr.21: 36461494-36461494 on Build GRCh38
rs922801470 Chr.21: 36461446-36461446 on Build GRCh38
rs757334760 Chr.21: 36461515-36461515 on Build GRCh38
rs765103185 Chr.21: 36461528-36461529 on Build GRCh38
rs74315438 Chr.21: 36461395-36461395 on Build GRCh38
rs975450328 Chr.21: 36461445-36461445 on Build GRCh38
rs748110915 Chr.21: 36461382-36461382 on Build GRCh38
rs373823945 Chr.21: 36461373-36461373 on Build GRCh38
rs747254086 Chr.21: 36461379-36461379 on Build GRCh38
rs756095919 Chr.21: 36461493-36461493 on Build GRCh38
rs781318012 Chr.21: 36461428-36461428 on Build GRCh38
rs763489263 Chr.21: 36461478-36461478 on Build GRCh38
rs752992781 Chr.21: 36461488-36461488 on Build GRCh38
rs748115585 Chr.21: 36461429-36461429 on Build GRCh38
rs371106978 Chr.21: 36461401-36461401 on Build GRCh38
rs370315827 Chr.21: 36461480-36461480 on Build GRCh38
rs560052879 Chr.21: 36461473-36461473 on Build GRCh38

View Full Product Details