Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73120376-73120649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ5
Entrez Gene ID
56479
Gene Name
potassium voltage-gated channel subfamily Q member 5
Gene Aliases
Kv7.5
UniGene
Hs.445324
Gene Chromosome Location
Chr.6: 72621843-73198851 on Build GRCh38
Transcripts
NM_001160130.1 NM_001160132.1 NM_001160133.1 NM_001160134.1 NM_019842.3 XM_017011058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654134 c.1186C>T p.H396Y Chr.6: 73120543-73120543 on Build GRCh38
3162125 c.1136G>A p.R379H Chr.6: 73120493-73120493 on Build GRCh38
4356858 c.1136G>A p.R379H Chr.6: 73120493-73120493 on Build GRCh38
7151698 c.1136G>A p.R379H Chr.6: 73120493-73120493 on Build GRCh38
4356857 c.1136G>A p.R379H Chr.6: 73120493-73120493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748453298 Chr.6: 73120596-73120596 on Build GRCh38
rs769020698 Chr.6: 73120456-73120456 on Build GRCh38
rs757962737 Chr.6: 73120583-73120583 on Build GRCh38
rs144427126 Chr.6: 73120500-73120500 on Build GRCh38
rs765646540 Chr.6: 73120488-73120488 on Build GRCh38
rs768203520 Chr.6: 73120592-73120592 on Build GRCh38
rs751731456 Chr.6: 73120497-73120497 on Build GRCh38
rs200362919 Chr.6: 73120526-73120526 on Build GRCh38
rs148067463 Chr.6: 73120560-73120560 on Build GRCh38
rs773773458 Chr.6: 73120492-73120492 on Build GRCh38
rs969180848 Chr.6: 73120608-73120608 on Build GRCh38
rs745844154 Chr.6: 73120574-73120574 on Build GRCh38
rs944048965 Chr.6: 73120423-73120423 on Build GRCh38
rs1008483524 Chr.6: 73120471-73120471 on Build GRCh38
rs746636980 Chr.6: 73120590-73120590 on Build GRCh38
rs776860106 Chr.6: 73120461-73120461 on Build GRCh38
rs757135342 Chr.6: 73120545-73120545 on Build GRCh38
rs1041269793 Chr.6: 73120472-73120472 on Build GRCh38
rs745360493 Chr.6: 73120442-73120442 on Build GRCh38
rs73753261 Chr.6: 73120450-73120450 on Build GRCh38
rs778020510 Chr.6: 73120441-73120441 on Build GRCh38
rs899854714 Chr.6: 73120462-73120462 on Build GRCh38
rs1037440994 Chr.6: 73120417-73120417 on Build GRCh38
rs577164847 Chr.6: 73120493-73120493 on Build GRCh38
rs779736928 Chr.6: 73120589-73120589 on Build GRCh38
rs762319678 Chr.6: 73120477-73120477 on Build GRCh38
rs766493000 Chr.6: 73120494-73120494 on Build GRCh38
rs757966813 Chr.6: 73120586-73120586 on Build GRCh38
rs890044336 Chr.6: 73120469-73120469 on Build GRCh38
rs746712024 Chr.6: 73120451-73120451 on Build GRCh38
rs760213630 Chr.6: 73120444-73120445 on Build GRCh38
rs369127759 Chr.6: 73120439-73120439 on Build GRCh38
rs544418404 Chr.6: 73120503-73120503 on Build GRCh38
rs534453813 Chr.6: 73120422-73120422 on Build GRCh38
rs770231936 Chr.6: 73120602-73120602 on Build GRCh38
rs780952561 Chr.6: 73120595-73120595 on Build GRCh38

View Full Product Details