Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148483102-148483373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
UniGene
Hs.483773
Gene Chromosome Location
Chr.5: 148451032-148654527 on Build GRCh38
Transcripts
NM_000870.6 NM_001040169.2 NM_001040172.2 NM_001040173.2 NM_001286410.1 NM_199453.3 NR_104445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4423449 c.1163C>T p.P388L Chr.5: 148483249-148483249 on Build GRCh38
4423448 c.1241C>T p.P414L Chr.5: 148483249-148483249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374062446 Chr.5: 148483169-148483169 on Build GRCh38
rs371669491 Chr.5: 148483248-148483248 on Build GRCh38
rs573735780 Chr.5: 148483127-148483127 on Build GRCh38
rs34826744 Chr.5: 148483255-148483255 on Build GRCh38
rs772811785 Chr.5: 148483305-148483307 on Build GRCh38
rs936334333 Chr.5: 148483301-148483301 on Build GRCh38
rs747696908 Chr.5: 148483295-148483296 on Build GRCh38
rs200903294 Chr.5: 148483323-148483323 on Build GRCh38
rs776602296 Chr.5: 148483246-148483246 on Build GRCh38
rs1036209488 Chr.5: 148483179-148483179 on Build GRCh38
rs764756332 Chr.5: 148483252-148483252 on Build GRCh38
rs778159489 Chr.5: 148483195-148483195 on Build GRCh38
rs757613271 Chr.5: 148483333-148483333 on Build GRCh38
rs201836791 Chr.5: 148483288-148483288 on Build GRCh38
rs201432293 Chr.5: 148483280-148483280 on Build GRCh38
rs756193511 Chr.5: 148483189-148483189 on Build GRCh38
rs201533186 Chr.5: 148483281-148483281 on Build GRCh38
rs772000556 Chr.5: 148483213-148483213 on Build GRCh38
rs546666430 Chr.5: 148483303-148483303 on Build GRCh38
rs200215537 Chr.5: 148483129-148483129 on Build GRCh38
rs201438004 Chr.5: 148483327-148483327 on Build GRCh38
rs779275477 Chr.5: 148483334-148483334 on Build GRCh38
rs749484929 Chr.5: 148483202-148483202 on Build GRCh38
rs893959291 Chr.5: 148483348-148483348 on Build GRCh38
rs565538004 Chr.5: 148483301-148483303 on Build GRCh38
rs199568633 Chr.5: 148483292-148483292 on Build GRCh38
rs932951154 Chr.5: 148483220-148483220 on Build GRCh38
rs1006132592 Chr.5: 148483249-148483249 on Build GRCh38
rs768545026 Chr.5: 148483245-148483245 on Build GRCh38
rs201813575 Chr.5: 148483241-148483241 on Build GRCh38
rs553152961 Chr.5: 148483134-148483134 on Build GRCh38
rs767954442 Chr.5: 148483284-148483284 on Build GRCh38
rs773954140 Chr.5: 148483231-148483231 on Build GRCh38
rs775370855 Chr.5: 148483231-148483231 on Build GRCh38
rs376772791 Chr.5: 148483336-148483336 on Build GRCh38
rs886529948 Chr.5: 148483146-148483146 on Build GRCh38
rs376816281 Chr.5: 148483272-148483272 on Build GRCh38
rs1003303295 Chr.5: 148483141-148483141 on Build GRCh38
rs201415979 Chr.5: 148483156-148483156 on Build GRCh38
rs755335782 Chr.5: 148483282-148483282 on Build GRCh38
rs1016352831 Chr.5: 148483191-148483191 on Build GRCh38
rs201253747 Chr.5: 148483142-148483142 on Build GRCh38

View Full Product Details