Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123059915-123060150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1559342 c.677C>A p.T226N Chr.11: 123059916-123059916 on Build GRCh38
304754 c.566T>G p.V189G Chr.11: 123060027-123060027 on Build GRCh38
924152 c.648C>A p.I216I Chr.11: 123059945-123059945 on Build GRCh38
6338891 c.639G>A p.E213E Chr.11: 123059954-123059954 on Build GRCh38
7154147 c.652G>C p.E218Q Chr.11: 123059941-123059941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200486073 Chr.11: 123060026-123060026 on Build GRCh38
rs775427523 Chr.11: 123060021-123060021 on Build GRCh38
rs763568867 Chr.11: 123060098-123060098 on Build GRCh38
rs573467766 Chr.11: 123059999-123059999 on Build GRCh38
rs764537309 Chr.11: 123060044-123060044 on Build GRCh38
rs370535447 Chr.11: 123060063-123060063 on Build GRCh38
rs199546915 Chr.11: 123060048-123060048 on Build GRCh38
rs199619043 Chr.11: 123060027-123060027 on Build GRCh38
rs749654902 Chr.11: 123060005-123060005 on Build GRCh38
rs894710340 Chr.11: 123060078-123060078 on Build GRCh38
rs753317621 Chr.11: 123059936-123059936 on Build GRCh38
rs906162427 Chr.11: 123060107-123060107 on Build GRCh38
rs41297901 Chr.11: 123060065-123060065 on Build GRCh38
rs181187850 Chr.11: 123060052-123060052 on Build GRCh38
rs376864898 Chr.11: 123060038-123060038 on Build GRCh38
rs373796555 Chr.11: 123060035-123060035 on Build GRCh38
rs1013205195 Chr.11: 123060089-123060089 on Build GRCh38
rs368508401 Chr.11: 123060090-123060090 on Build GRCh38
rs770919426 Chr.11: 123060077-123060077 on Build GRCh38
rs200266481 Chr.11: 123060045-123060045 on Build GRCh38
rs951965347 Chr.11: 123059987-123059987 on Build GRCh38
rs562265344 Chr.11: 123060093-123060093 on Build GRCh38
rs376403473 Chr.11: 123059996-123059996 on Build GRCh38
rs756568266 Chr.11: 123059960-123059960 on Build GRCh38
rs751301386 Chr.11: 123060033-123060033 on Build GRCh38
rs766545520 Chr.11: 123060094-123060094 on Build GRCh38
rs772233016 Chr.11: 123060020-123060020 on Build GRCh38
rs775639640 Chr.11: 123060042-123060042 on Build GRCh38
rs760868592 Chr.11: 123060069-123060073 on Build GRCh38
rs773246178 Chr.11: 123060023-123060023 on Build GRCh38
rs764390903 Chr.11: 123060079-123060083 on Build GRCh38
rs778283472 Chr.11: 123059968-123059968 on Build GRCh38
rs746155109 Chr.11: 123060049-123060049 on Build GRCh38
rs757663783 Chr.11: 123060046-123060046 on Build GRCh38
rs577056442 Chr.11: 123060061-123060061 on Build GRCh38
rs769624839 Chr.11: 123060022-123060022 on Build GRCh38
rs780263669 Chr.11: 123060054-123060054 on Build GRCh38
rs779126635 Chr.11: 123060011-123060011 on Build GRCh38
rs1041755663 Chr.11: 123060060-123060060 on Build GRCh38
rs762901745 Chr.11: 123060034-123060034 on Build GRCh38
rs774277159 Chr.11: 123060087-123060087 on Build GRCh38
rs944711644 Chr.11: 123060053-123060053 on Build GRCh38

View Full Product Details