Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49889300-49889573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376432 c.647C>T p.T216I Chr.12: 49889485-49889485 on Build GRCh38
4754963 c.583G>A p.V195M Chr.12: 49889549-49889549 on Build GRCh38
7162642 c.630C>T p.T210T Chr.12: 49889502-49889502 on Build GRCh38
4943676 c.576C>T p.T192T Chr.12: 49889556-49889556 on Build GRCh38
1362196 c.621C>G p.L207L Chr.12: 49889511-49889511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758363486 Chr.12: 49889453-49889453 on Build GRCh38
rs941845658 Chr.12: 49889417-49889417 on Build GRCh38
rs535057833 Chr.12: 49889341-49889341 on Build GRCh38
rs888870729 Chr.12: 49889416-49889416 on Build GRCh38
rs150732432 Chr.12: 49889501-49889501 on Build GRCh38
rs776551275 Chr.12: 49889434-49889434 on Build GRCh38
rs546605807 Chr.12: 49889327-49889327 on Build GRCh38
rs756424544 Chr.12: 49889467-49889467 on Build GRCh38
rs961934759 Chr.12: 49889374-49889374 on Build GRCh38
rs746490841 Chr.12: 49889503-49889503 on Build GRCh38
rs749469222 Chr.12: 49889480-49889480 on Build GRCh38
rs80246888 Chr.12: 49889524-49889524 on Build GRCh38
rs763025973 Chr.12: 49889550-49889550 on Build GRCh38
rs933005704 Chr.12: 49889404-49889404 on Build GRCh38
rs187052883 Chr.12: 49889487-49889487 on Build GRCh38
rs375275250 Chr.12: 49889511-49889511 on Build GRCh38
rs74953115 Chr.12: 49889395-49889395 on Build GRCh38
rs185815389 Chr.12: 49889320-49889320 on Build GRCh38
rs759267308 Chr.12: 49889443-49889443 on Build GRCh38
rs765486396 Chr.12: 49889486-49889486 on Build GRCh38
rs1037925997 Chr.12: 49889463-49889463 on Build GRCh38
rs995657751 Chr.12: 49889382-49889383 on Build GRCh38
rs989219867 Chr.12: 49889422-49889422 on Build GRCh38
rs181154832 Chr.12: 49889412-49889412 on Build GRCh38
rs750437175 Chr.12: 49889458-49889458 on Build GRCh38
rs80295142 Chr.12: 49889332-49889332 on Build GRCh38
rs139044559 Chr.12: 49889502-49889502 on Build GRCh38
rs764121293 Chr.12: 49889551-49889551 on Build GRCh38
rs776157297 Chr.12: 49889523-49889523 on Build GRCh38
rs970221698 Chr.12: 49889396-49889396 on Build GRCh38
rs189787605 Chr.12: 49889367-49889367 on Build GRCh38
rs759853593 Chr.12: 49889398-49889398 on Build GRCh38
rs748673361 Chr.12: 49889493-49889493 on Build GRCh38
rs775403937 Chr.12: 49889549-49889549 on Build GRCh38
rs765147386 Chr.12: 49889526-49889526 on Build GRCh38
rs765057686 Chr.12: 49889452-49889452 on Build GRCh38

View Full Product Details