Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48966576-48966835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTL
Entrez Gene ID
2512
Gene Name
ferritin light chain
Gene Aliases
LFTD, NBIA3
UniGene
Hs.433670
Gene Chromosome Location
Chr.19: 48965309-48966879 on Build GRCh38
Transcripts
NM_000146.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761476 c.376-2A>C p.? Chr.19: 48966581-48966581 on Build GRCh38
7212433 c.408T>G p.D136E Chr.19: 48966615-48966615 on Build GRCh38
3835334 c.501C>T p.F167F Chr.19: 48966708-48966708 on Build GRCh38
3536713 c.449C>T p.T150I Chr.19: 48966656-48966656 on Build GRCh38
439938 c.382G>A p.D128N Chr.19: 48966589-48966589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760853527 Chr.19: 48966727-48966727 on Build GRCh38
rs778872543 Chr.19: 48966677-48966677 on Build GRCh38
rs981387168 Chr.19: 48966607-48966607 on Build GRCh38
rs11553249 Chr.19: 48966767-48966767 on Build GRCh38
rs11553236 Chr.19: 48966715-48966715 on Build GRCh38
rs398124640 Chr.19: 48966691-48966692 on Build GRCh38
rs111962558 Chr.19: 48966643-48966643 on Build GRCh38
rs571937803 Chr.19: 48966770-48966770 on Build GRCh38
rs536646625 Chr.19: 48966807-48966807 on Build GRCh38
rs3211586 Chr.19: 48966739-48966739 on Build GRCh38
rs11553216 Chr.19: 48966649-48966649 on Build GRCh38
rs758414077 Chr.19: 48966681-48966681 on Build GRCh38
rs755825625 Chr.19: 48966656-48966656 on Build GRCh38
rs753909441 Chr.19: 48966743-48966743 on Build GRCh38
rs11553250 Chr.19: 48966740-48966740 on Build GRCh38
rs766011614 Chr.19: 48966719-48966720 on Build GRCh38
rs900999430 Chr.19: 48966734-48966734 on Build GRCh38
rs779807848 Chr.19: 48966682-48966682 on Build GRCh38
rs370329863 Chr.19: 48966678-48966678 on Build GRCh38
rs11553214 Chr.19: 48966664-48966664 on Build GRCh38
rs778845273 Chr.19: 48966801-48966801 on Build GRCh38
rs764635265 Chr.19: 48966753-48966753 on Build GRCh38
rs939447629 Chr.19: 48966661-48966661 on Build GRCh38
rs769933942 Chr.19: 48966693-48966693 on Build GRCh38
rs1052502825 Chr.19: 48966672-48966672 on Build GRCh38
rs1038871067 Chr.19: 48966640-48966640 on Build GRCh38
rs3211589 Chr.19: 48966749-48966749 on Build GRCh38
rs530889896 Chr.19: 48966632-48966632 on Build GRCh38
rs373178636 Chr.19: 48966743-48966743 on Build GRCh38
rs753342915 Chr.19: 48966670-48966670 on Build GRCh38
rs779967554 Chr.19: 48966768-48966768 on Build GRCh38
rs138091847 Chr.19: 48966782-48966782 on Build GRCh38
rs73046709 Chr.19: 48966729-48966729 on Build GRCh38
rs151265703 Chr.19: 48966673-48966673 on Build GRCh38
rs3211587 Chr.19: 48966742-48966742 on Build GRCh38
rs1803578 Chr.19: 48966727-48966727 on Build GRCh38
rs774740467 Chr.19: 48966609-48966609 on Build GRCh38
rs771233352 Chr.19: 48966787-48966787 on Build GRCh38
rs551215005 Chr.19: 48966802-48966802 on Build GRCh38
rs777930451 Chr.19: 48966786-48966786 on Build GRCh38
rs1803580 Chr.19: 48966721-48966721 on Build GRCh38
rs770946603 Chr.19: 48966699-48966699 on Build GRCh38
rs774529369 Chr.19: 48966703-48966703 on Build GRCh38
rs762943867 Chr.19: 48966606-48966606 on Build GRCh38
rs374486686 Chr.19: 48966680-48966680 on Build GRCh38
rs368083362 Chr.19: 48966759-48966759 on Build GRCh38
rs746870331 Chr.19: 48966691-48966691 on Build GRCh38
rs763791960 Chr.19: 48966737-48966737 on Build GRCh38
rs768204975 Chr.19: 48966709-48966709 on Build GRCh38
rs569313546 Chr.19: 48966803-48966803 on Build GRCh38
rs776164311 Chr.19: 48966804-48966804 on Build GRCh38
rs951133608 Chr.19: 48966754-48966754 on Build GRCh38
rs759937926 Chr.19: 48966708-48966708 on Build GRCh38
rs764013805 Chr.19: 48966766-48966766 on Build GRCh38
rs3211588 Chr.19: 48966746-48966746 on Build GRCh38
rs756977598 Chr.19: 48966751-48966751 on Build GRCh38
rs759903976 Chr.19: 48966616-48966616 on Build GRCh38
rs11553270 Chr.19: 48966717-48966717 on Build GRCh38
rs773434208 Chr.19: 48966696-48966696 on Build GRCh38
rs587776840 Chr.19: 48966665-48966666 on Build GRCh38
rs368634782 Chr.19: 48966698-48966698 on Build GRCh38
rs11553198 Chr.19: 48966684-48966684 on Build GRCh38
rs754491772 Chr.19: 48966766-48966772 on Build GRCh38
rs560486159 Chr.19: 48966761-48966761 on Build GRCh38

View Full Product Details