Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49946042-49946315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB133
Entrez Gene ID
403339
Gene Name
defensin beta 133
Gene Aliases
-
UniGene
Hs.558270
Gene Chromosome Location
Chr.6: 49946101-49949444 on Build GRCh38
Transcripts
NM_001166478.1 XM_017010860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239887 c.174C>A p.N58K Chr.6: 49946113-49946113 on Build GRCh38
1080104 c.93C>T p.C31C Chr.6: 49946194-49946194 on Build GRCh38
1203320 c.115C>T p.R39* Chr.6: 49946172-49946172 on Build GRCh38
1203321 c.153C>A p.F51L Chr.6: 49946134-49946134 on Build GRCh38
6858649 c.89G>C p.S30T Chr.6: 49946198-49946198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145585477 Chr.6: 49946124-49946124 on Build GRCh38
rs766667615 Chr.6: 49946215-49946215 on Build GRCh38
rs957150223 Chr.6: 49946260-49946260 on Build GRCh38
rs149691409 Chr.6: 49946086-49946086 on Build GRCh38
rs142977787 Chr.6: 49946144-49946145 on Build GRCh38
rs775275031 Chr.6: 49946093-49946093 on Build GRCh38
rs368968597 Chr.6: 49946207-49946207 on Build GRCh38
rs977113007 Chr.6: 49946223-49946223 on Build GRCh38
rs560130001 Chr.6: 49946174-49946174 on Build GRCh38
rs745516514 Chr.6: 49946082-49946082 on Build GRCh38
rs7760672 Chr.6: 49946268-49946268 on Build GRCh38
rs907234023 Chr.6: 49946159-49946159 on Build GRCh38
rs936207170 Chr.6: 49946088-49946088 on Build GRCh38
rs948986357 Chr.6: 49946129-49946129 on Build GRCh38
rs17664085 Chr.6: 49946214-49946214 on Build GRCh38
rs1004201964 Chr.6: 49946184-49946184 on Build GRCh38
rs1055753519 Chr.6: 49946085-49946085 on Build GRCh38
rs751762339 Chr.6: 49946216-49946216 on Build GRCh38
rs995963738 Chr.6: 49946203-49946203 on Build GRCh38
rs532278129 Chr.6: 49946270-49946270 on Build GRCh38
rs965737606 Chr.6: 49946217-49946217 on Build GRCh38
rs114979966 Chr.6: 49946077-49946077 on Build GRCh38
rs768547767 Chr.6: 49946166-49946166 on Build GRCh38
rs190254963 Chr.6: 49946158-49946158 on Build GRCh38
rs920068390 Chr.6: 49946235-49946235 on Build GRCh38
rs1037555234 Chr.6: 49946189-49946189 on Build GRCh38
rs776731408 Chr.6: 49946179-49946179 on Build GRCh38
rs761775480 Chr.6: 49946183-49946183 on Build GRCh38
rs898584215 Chr.6: 49946198-49946198 on Build GRCh38
rs35647105 Chr.6: 49946107-49946108 on Build GRCh38
rs1031713310 Chr.6: 49946226-49946226 on Build GRCh38

View Full Product Details