Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89149233-89149488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP7
Entrez Gene ID
388646
Gene Name
guanylate binding protein 7
Gene Aliases
GBP4L
UniGene
Hs.591450
Gene Chromosome Location
Chr.1: 89131751-89176040 on Build GRCh38
Transcripts
NM_207398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
128052 c.1026G>A p.V342V Chr.1: 89149418-89149418 on Build GRCh38
4686564 c.1094C>T p.A365V Chr.1: 89149350-89149350 on Build GRCh38
3790323 c.989C>T p.A330V Chr.1: 89149455-89149455 on Build GRCh38
4686565 c.1062C>T p.D354D Chr.1: 89149382-89149382 on Build GRCh38
3493165 c.1138C>T p.Q380* Chr.1: 89149306-89149306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755181722 Chr.1: 89149300-89149300 on Build GRCh38
rs535795589 Chr.1: 89149435-89149435 on Build GRCh38
rs766750851 Chr.1: 89149268-89149268 on Build GRCh38
rs767570581 Chr.1: 89149353-89149353 on Build GRCh38
rs201056728 Chr.1: 89149341-89149341 on Build GRCh38
rs946315860 Chr.1: 89149392-89149392 on Build GRCh38
rs369410469 Chr.1: 89149281-89149281 on Build GRCh38
rs74098339 Chr.1: 89149340-89149340 on Build GRCh38
rs996355331 Chr.1: 89149464-89149464 on Build GRCh38
rs779341002 Chr.1: 89149312-89149312 on Build GRCh38
rs560981037 Chr.1: 89149381-89149381 on Build GRCh38
rs571395404 Chr.1: 89149276-89149276 on Build GRCh38
rs757088922 Chr.1: 89149352-89149352 on Build GRCh38
rs372954977 Chr.1: 89149296-89149296 on Build GRCh38
rs749396805 Chr.1: 89149298-89149298 on Build GRCh38
rs754456593 Chr.1: 89149268-89149269 on Build GRCh38
rs747414013 Chr.1: 89149384-89149384 on Build GRCh38
rs143684763 Chr.1: 89149424-89149424 on Build GRCh38
rs779425164 Chr.1: 89149267-89149268 on Build GRCh38
rs757252155 Chr.1: 89149293-89149293 on Build GRCh38
rs745362797 Chr.1: 89149426-89149426 on Build GRCh38
rs199576629 Chr.1: 89149422-89149422 on Build GRCh38
rs759139135 Chr.1: 89149275-89149275 on Build GRCh38
rs776148123 Chr.1: 89149268-89149268 on Build GRCh38
rs752484582 Chr.1: 89149279-89149279 on Build GRCh38
rs765845130 Chr.1: 89149261-89149261 on Build GRCh38
rs773736455 Chr.1: 89149320-89149320 on Build GRCh38
rs776060427 Chr.1: 89149329-89149329 on Build GRCh38
rs947803358 Chr.1: 89149318-89149318 on Build GRCh38
rs775015333 Chr.1: 89149463-89149463 on Build GRCh38
rs769335151 Chr.1: 89149447-89149447 on Build GRCh38
rs751494980 Chr.1: 89149289-89149289 on Build GRCh38
rs952048498 Chr.1: 89149262-89149262 on Build GRCh38
rs748355172 Chr.1: 89149363-89149363 on Build GRCh38
rs200457249 Chr.1: 89149323-89149323 on Build GRCh38
rs142621996 Chr.1: 89149317-89149317 on Build GRCh38
rs753959935 Chr.1: 89149354-89149354 on Build GRCh38
rs771339090 Chr.1: 89149390-89149390 on Build GRCh38
rs755163316 Chr.1: 89149355-89149355 on Build GRCh38
rs542953952 Chr.1: 89149347-89149347 on Build GRCh38
rs570295547 Chr.1: 89149283-89149283 on Build GRCh38
rs891522595 Chr.1: 89149327-89149327 on Build GRCh38
rs531634408 Chr.1: 89149465-89149465 on Build GRCh38
rs1040562121 Chr.1: 89149393-89149393 on Build GRCh38
rs779252964 Chr.1: 89149361-89149361 on Build GRCh38
rs777949392 Chr.1: 89149382-89149382 on Build GRCh38
rs577448421 Chr.1: 89149316-89149316 on Build GRCh38
rs145429337 Chr.1: 89149322-89149322 on Build GRCh38
rs763759581 Chr.1: 89149342-89149342 on Build GRCh38
rs759114440 Chr.1: 89149335-89149335 on Build GRCh38
rs768372837 Chr.1: 89149466-89149466 on Build GRCh38

View Full Product Details