Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91056130-91056383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
UniGene
Hs.125750
Gene Chromosome Location
Chr.12: 91050491-91058354 on Build GRCh38
Transcripts
NM_007035.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940282 c.91G>A p.D31N Chr.12: 91056191-91056191 on Build GRCh38
2044947 c.96A>G p.S32S Chr.12: 91056186-91056186 on Build GRCh38
3813188 c.81T>C p.Y27Y Chr.12: 91056201-91056201 on Build GRCh38
240335 c.22A>G p.I8V Chr.12: 91056260-91056260 on Build GRCh38
5551330 c.16T>C p.C6R Chr.12: 91056266-91056266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749186513 Chr.12: 91056176-91056176 on Build GRCh38
rs749435376 Chr.12: 91056198-91056198 on Build GRCh38
rs780006810 Chr.12: 91056275-91056275 on Build GRCh38
rs201524750 Chr.12: 91056196-91056196 on Build GRCh38
rs752745106 Chr.12: 91056257-91056257 on Build GRCh38
rs1026475863 Chr.12: 91056216-91056216 on Build GRCh38
rs1057060712 Chr.12: 91056327-91056327 on Build GRCh38
rs370775164 Chr.12: 91056180-91056180 on Build GRCh38
rs1009131291 Chr.12: 91056233-91056233 on Build GRCh38
rs755201083 Chr.12: 91056251-91056251 on Build GRCh38
rs746591463 Chr.12: 91056277-91056277 on Build GRCh38
rs757391405 Chr.12: 91056226-91056227 on Build GRCh38
rs750443030 Chr.12: 91056273-91056273 on Build GRCh38
rs751725649 Chr.12: 91056242-91056242 on Build GRCh38
rs773814068 Chr.12: 91056226-91056226 on Build GRCh38
rs747586192 Chr.12: 91056316-91056316 on Build GRCh38
rs968432306 Chr.12: 91056281-91056281 on Build GRCh38
rs543283787 Chr.12: 91056349-91056349 on Build GRCh38
rs12320366 Chr.12: 91056207-91056207 on Build GRCh38
rs775278959 Chr.12: 91056200-91056200 on Build GRCh38
rs758255217 Chr.12: 91056274-91056274 on Build GRCh38
rs2735333 Chr.12: 91056213-91056213 on Build GRCh38
rs778721242 Chr.12: 91056186-91056186 on Build GRCh38
rs751817390 Chr.12: 91056161-91056161 on Build GRCh38
rs762340226 Chr.12: 91056221-91056221 on Build GRCh38
rs202094304 Chr.12: 91056154-91056154 on Build GRCh38
rs765681497 Chr.12: 91056223-91056223 on Build GRCh38
rs147494238 Chr.12: 91056264-91056264 on Build GRCh38
rs773584773 Chr.12: 91056326-91056326 on Build GRCh38
rs569894609 Chr.12: 91056193-91056193 on Build GRCh38
rs769382533 Chr.12: 91056325-91056325 on Build GRCh38
rs866472061 Chr.12: 91056259-91056259 on Build GRCh38
rs758594193 Chr.12: 91056151-91056151 on Build GRCh38
rs763505751 Chr.12: 91056332-91056332 on Build GRCh38
rs767549485 Chr.12: 91056256-91056256 on Build GRCh38
rs756267298 Chr.12: 91056261-91056261 on Build GRCh38
rs763193954 Chr.12: 91056227-91056227 on Build GRCh38
rs141931395 Chr.12: 91056283-91056283 on Build GRCh38
rs28776777 Chr.12: 91056305-91056305 on Build GRCh38
rs199813838 Chr.12: 91056162-91056162 on Build GRCh38
rs377616201 Chr.12: 91056172-91056172 on Build GRCh38
rs778947165 Chr.12: 91056296-91056296 on Build GRCh38
rs766793375 Chr.12: 91056231-91056231 on Build GRCh38
rs746385866 Chr.12: 91056313-91056314 on Build GRCh38

View Full Product Details