Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51064558-51064809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP2
Entrez Gene ID
81566
Gene Name
cysteine and serine rich nuclear protein 2
Gene Aliases
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12
UniGene
Hs.524425
Gene Chromosome Location
Chr.12: 51061205-51083671 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

Gene Symbol
LETMD1
Entrez Gene ID
25875
Gene Name
LETM1 domain containing 1
Gene Aliases
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
UniGene
Hs.655272
Gene Chromosome Location
Chr.12: 51048230-51068984 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5841200 c.791G>A p.R264Q Chr.12: 51064587-51064587 on Build GRCh38
6227497 c.769C>T p.R257C Chr.12: 51064609-51064609 on Build GRCh38
4508851 c.789C>T p.I263I Chr.12: 51064589-51064589 on Build GRCh38
6072774 c.818T>G p.I273S Chr.12: 51064560-51064560 on Build GRCh38
4558087 c.754G>A p.G252R Chr.12: 51064624-51064624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77607059 Chr.12: 51064621-51064621 on Build GRCh38
rs900227274 Chr.12: 51064692-51064692 on Build GRCh38
rs377430706 Chr.12: 51064699-51064699 on Build GRCh38
rs11832566 Chr.12: 51064719-51064719 on Build GRCh38
rs148790966 Chr.12: 51064672-51064672 on Build GRCh38
rs370522473 Chr.12: 51064703-51064703 on Build GRCh38
rs888726374 Chr.12: 51064728-51064728 on Build GRCh38
rs1005784545 Chr.12: 51064730-51064730 on Build GRCh38
rs1039826502 Chr.12: 51064694-51064694 on Build GRCh38
rs770856679 Chr.12: 51064744-51064744 on Build GRCh38
rs767016607 Chr.12: 51064632-51064632 on Build GRCh38
rs751091442 Chr.12: 51064683-51064683 on Build GRCh38
rs777016214 Chr.12: 51064636-51064636 on Build GRCh38
rs748314697 Chr.12: 51064710-51064710 on Build GRCh38
rs866368495 Chr.12: 51064737-51064737 on Build GRCh38
rs761211045 Chr.12: 51064631-51064631 on Build GRCh38
rs2029147 Chr.12: 51064775-51064775 on Build GRCh38
rs773434648 Chr.12: 51064605-51064605 on Build GRCh38
rs778852688 Chr.12: 51064777-51064779 on Build GRCh38
rs1051238154 Chr.12: 51064732-51064732 on Build GRCh38
rs376930680 Chr.12: 51064649-51064649 on Build GRCh38
rs4768957 Chr.12: 51064676-51064676 on Build GRCh38
rs142481237 Chr.12: 51064769-51064769 on Build GRCh38
rs779968251 Chr.12: 51064707-51064707 on Build GRCh38
rs758047907 Chr.12: 51064675-51064675 on Build GRCh38
rs779342595 Chr.12: 51064680-51064682 on Build GRCh38
rs747120797 Chr.12: 51064720-51064720 on Build GRCh38
rs765689102 Chr.12: 51064646-51064646 on Build GRCh38
rs560057159 Chr.12: 51064591-51064591 on Build GRCh38
rs573755995 Chr.12: 51064640-51064640 on Build GRCh38
rs772276603 Chr.12: 51064582-51064582 on Build GRCh38
rs373134162 Chr.12: 51064688-51064688 on Build GRCh38
rs11169692 Chr.12: 51064782-51064782 on Build GRCh38
rs1004564902 Chr.12: 51064762-51064762 on Build GRCh38

View Full Product Details