Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237097478-237097751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS8
Entrez Gene ID
10920
Gene Name
COP9 signalosome subunit 8
Gene Aliases
COP9, CSN8, SGN8
UniGene
Hs.531713
Gene Chromosome Location
Chr.2: 237085441-237098846 on Build GRCh38
Transcripts
NM_006710.4 NM_198189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301737 c.561A>T p.P187P Chr.2: 237097673-237097673 on Build GRCh38
1202083 c.553C>T p.P185S Chr.2: 237097665-237097665 on Build GRCh38
4500251 c.560C>T p.P187L Chr.2: 237097672-237097672 on Build GRCh38
7099960 c.622G>C p.E208Q Chr.2: 237097734-237097734 on Build GRCh38
3051221 c.570A>G p.P190P Chr.2: 237097682-237097682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571245706 Chr.2: 237097524-237097524 on Build GRCh38
rs1034559868 Chr.2: 237097566-237097566 on Build GRCh38
rs761257535 Chr.2: 237097669-237097669 on Build GRCh38
rs1032880423 Chr.2: 237097525-237097525 on Build GRCh38
rs889554376 Chr.2: 237097598-237097598 on Build GRCh38
rs776869813 Chr.2: 237097619-237097619 on Build GRCh38
rs959043582 Chr.2: 237097585-237097586 on Build GRCh38
rs557901190 Chr.2: 237097720-237097720 on Build GRCh38
rs763948366 Chr.2: 237097644-237097644 on Build GRCh38
rs536018614 Chr.2: 237097588-237097588 on Build GRCh38
rs34606518 Chr.2: 237097714-237097715 on Build GRCh38
rs535306075 Chr.2: 237097624-237097624 on Build GRCh38
rs185500850 Chr.2: 237097556-237097556 on Build GRCh38
rs779489336 Chr.2: 237097683-237097683 on Build GRCh38
rs368533932 Chr.2: 237097682-237097682 on Build GRCh38
rs959466631 Chr.2: 237097539-237097539 on Build GRCh38
rs72975809 Chr.2: 237097586-237097586 on Build GRCh38
rs753488393 Chr.2: 237097652-237097652 on Build GRCh38
rs771093474 Chr.2: 237097614-237097614 on Build GRCh38
rs189050402 Chr.2: 237097509-237097509 on Build GRCh38
rs377323601 Chr.2: 237097692-237097692 on Build GRCh38
rs767243543 Chr.2: 237097671-237097671 on Build GRCh38
rs750154058 Chr.2: 237097674-237097674 on Build GRCh38
rs972961083 Chr.2: 237097565-237097565 on Build GRCh38
rs181097361 Chr.2: 237097540-237097540 on Build GRCh38
rs1020490443 Chr.2: 237097554-237097554 on Build GRCh38
rs747071033 Chr.2: 237097713-237097713 on Build GRCh38
rs978610800 Chr.2: 237097507-237097507 on Build GRCh38
rs907749452 Chr.2: 237097673-237097673 on Build GRCh38
rs758884554 Chr.2: 237097699-237097699 on Build GRCh38
rs778166607 Chr.2: 237097705-237097705 on Build GRCh38
rs1009873765 Chr.2: 237097716-237097716 on Build GRCh38
rs890573104 Chr.2: 237097544-237097544 on Build GRCh38
rs373280338 Chr.2: 237097622-237097622 on Build GRCh38
rs761472807 Chr.2: 237097630-237097630 on Build GRCh38
rs775400197 Chr.2: 237097626-237097626 on Build GRCh38
rs900843141 Chr.2: 237097715-237097715 on Build GRCh38
rs762879954 Chr.2: 237097634-237097634 on Build GRCh38
rs901998982 Chr.2: 237097504-237097504 on Build GRCh38
rs1000455492 Chr.2: 237097560-237097560 on Build GRCh38
rs550668997 Chr.2: 237097551-237097551 on Build GRCh38
rs555921582 Chr.2: 237097595-237097595 on Build GRCh38
rs530878347 Chr.2: 237097659-237097659 on Build GRCh38

View Full Product Details