Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111694544-111694783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYN
Entrez Gene ID
2534
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Gene Aliases
SLK, SYN, p59-FYN
UniGene
Hs.390567
Gene Chromosome Location
Chr.6: 111660332-111873452 on Build GRCh38
Transcripts
NM_002037.5 NM_153047.3 NM_153048.3 XM_005266890.3 XM_005266892.3 XM_017010650.1 XM_017010651.1 XM_017010652.1 XM_017010653.1 XM_017010654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6883328 c.1041A>C p.L347F Chr.6: 111694697-111694697 on Build GRCh38
3410537 c.1078A>G p.K360E Chr.6: 111694660-111694660 on Build GRCh38
1487144 c.1041A>G p.L347L Chr.6: 111694697-111694697 on Build GRCh38
3725756 c.1081G>T p.A361S Chr.6: 111694666-111694666 on Build GRCh38
6928429 c.1069G>A p.G357R Chr.6: 111694678-111694678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778100987 Chr.6: 111694612-111694612 on Build GRCh38
rs746940763 Chr.6: 111694658-111694658 on Build GRCh38
rs869063474 Chr.6: 111694622-111694622 on Build GRCh38
rs768772210 Chr.6: 111694687-111694687 on Build GRCh38
rs753177376 Chr.6: 111694597-111694597 on Build GRCh38
rs897080714 Chr.6: 111694746-111694746 on Build GRCh38
rs776429653 Chr.6: 111694708-111694708 on Build GRCh38
rs371036122 Chr.6: 111694654-111694654 on Build GRCh38
rs559426859 Chr.6: 111694576-111694576 on Build GRCh38
rs804186 Chr.6: 111694620-111694620 on Build GRCh38
rs745335589 Chr.6: 111694631-111694631 on Build GRCh38
rs550786502 Chr.6: 111694585-111694585 on Build GRCh38
rs778465060 Chr.6: 111694622-111694622 on Build GRCh38
rs533306619 Chr.6: 111694733-111694733 on Build GRCh38
rs771687922 Chr.6: 111694640-111694640 on Build GRCh38
rs1040286611 Chr.6: 111694598-111694598 on Build GRCh38
rs759563917 Chr.6: 111694584-111694584 on Build GRCh38
rs374665881 Chr.6: 111694610-111694610 on Build GRCh38
rs1036974418 Chr.6: 111694665-111694665 on Build GRCh38
rs201834690 Chr.6: 111694578-111694578 on Build GRCh38
rs368283898 Chr.6: 111694754-111694754 on Build GRCh38

View Full Product Details