Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77928301-77928546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
UniGene
Hs.533723
Gene Chromosome Location
Chr.11: 77874491-77994849 on Build GRCh38
Transcripts
NM_033547.3 XM_011545352.2 XM_011545353.2 XM_017018560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5821677 c.1363G>T p.V455L Chr.11: 77928350-77928350 on Build GRCh38
5051157 c.1282C>T p.R428C Chr.11: 77928431-77928431 on Build GRCh38
6194381 c.1367T>C p.L456P Chr.11: 77928346-77928346 on Build GRCh38
5051156 c.1325T>C p.I442T Chr.11: 77928388-77928388 on Build GRCh38
7372330 c.1321A>C p.N441H Chr.11: 77928392-77928392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766703927 Chr.11: 77928482-77928482 on Build GRCh38
rs113532734 Chr.11: 77928336-77928336 on Build GRCh38
rs778939259 Chr.11: 77928395-77928396 on Build GRCh38
rs368475030 Chr.11: 77928405-77928405 on Build GRCh38
rs762497086 Chr.11: 77928456-77928456 on Build GRCh38
rs746126953 Chr.11: 77928441-77928441 on Build GRCh38
rs11607043 Chr.11: 77928420-77928420 on Build GRCh38
rs779006309 Chr.11: 77928397-77928397 on Build GRCh38
rs562652655 Chr.11: 77928515-77928515 on Build GRCh38
rs769847193 Chr.11: 77928438-77928438 on Build GRCh38
rs763814229 Chr.11: 77928459-77928459 on Build GRCh38
rs11607479 Chr.11: 77928465-77928465 on Build GRCh38
rs752219202 Chr.11: 77928351-77928351 on Build GRCh38
rs755456584 Chr.11: 77928395-77928395 on Build GRCh38
rs756369911 Chr.11: 77928385-77928385 on Build GRCh38
rs748223589 Chr.11: 77928398-77928398 on Build GRCh38
rs761126606 Chr.11: 77928478-77928478 on Build GRCh38
rs754171511 Chr.11: 77928490-77928490 on Build GRCh38
rs746851622 Chr.11: 77928430-77928430 on Build GRCh38
rs759583010 Chr.11: 77928437-77928437 on Build GRCh38
rs577201312 Chr.11: 77928389-77928389 on Build GRCh38
rs143540393 Chr.11: 77928392-77928392 on Build GRCh38
rs375319525 Chr.11: 77928356-77928356 on Build GRCh38
rs1016046185 Chr.11: 77928496-77928496 on Build GRCh38
rs556037618 Chr.11: 77928458-77928458 on Build GRCh38
rs554082325 Chr.11: 77928380-77928380 on Build GRCh38
rs146790163 Chr.11: 77928434-77928434 on Build GRCh38
rs775194729 Chr.11: 77928450-77928450 on Build GRCh38

View Full Product Details