Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95450068-95450341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81B
Entrez Gene ID
153643
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
UniGene
Hs.276287
Gene Chromosome Location
Chr.5: 95384793-95450441 on Build GRCh38
Transcripts
NM_152548.2 XM_005271905.2 XM_005271906.4 XM_011543209.1 XM_011543210.2 XM_017009118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950203 c.1333A>G p.K445E Chr.5: 95450256-95450256 on Build GRCh38
6399062 c.1235C>A p.S412* Chr.5: 95450158-95450158 on Build GRCh38
1071339 c.1229T>G p.F410C Chr.5: 95450152-95450152 on Build GRCh38
1251963 c.1349_1350insG p.E451fs*>3 Chr.5: 95450272-95450273 on Build GRCh38
70726 c.1284G>A p.M428I Chr.5: 95450207-95450207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747423587 Chr.5: 95450295-95450295 on Build GRCh38
rs771620475 Chr.5: 95450191-95450191 on Build GRCh38
rs199915561 Chr.5: 95450094-95450094 on Build GRCh38
rs117093563 Chr.5: 95450254-95450254 on Build GRCh38
rs369548823 Chr.5: 95450216-95450216 on Build GRCh38
rs761112863 Chr.5: 95450184-95450184 on Build GRCh38
rs950952710 Chr.5: 95450277-95450277 on Build GRCh38
rs773641186 Chr.5: 95450172-95450172 on Build GRCh38
rs778993021 Chr.5: 95450260-95450260 on Build GRCh38
rs763732152 Chr.5: 95450229-95450229 on Build GRCh38
rs751364470 Chr.5: 95450230-95450230 on Build GRCh38
rs373286610 Chr.5: 95450224-95450224 on Build GRCh38
rs573417076 Chr.5: 95450093-95450093 on Build GRCh38
rs751339959 Chr.5: 95450196-95450196 on Build GRCh38
rs867982926 Chr.5: 95450179-95450179 on Build GRCh38
rs201358737 Chr.5: 95450253-95450253 on Build GRCh38
rs768748899 Chr.5: 95450129-95450129 on Build GRCh38
rs771389119 Chr.5: 95450301-95450301 on Build GRCh38
rs562119047 Chr.5: 95450222-95450222 on Build GRCh38
rs377294269 Chr.5: 95450213-95450213 on Build GRCh38
rs754696634 Chr.5: 95450296-95450298 on Build GRCh38
rs779033228 Chr.5: 95450133-95450133 on Build GRCh38
rs748352054 Chr.5: 95450138-95450138 on Build GRCh38
rs778660851 Chr.5: 95450176-95450176 on Build GRCh38
rs375986616 Chr.5: 95450227-95450227 on Build GRCh38
rs192114736 Chr.5: 95450103-95450103 on Build GRCh38
rs751414261 Chr.5: 95450247-95450247 on Build GRCh38
rs764758376 Chr.5: 95450221-95450221 on Build GRCh38
rs766285774 Chr.5: 95450226-95450226 on Build GRCh38
rs778124684 Chr.5: 95450284-95450284 on Build GRCh38
rs1015738975 Chr.5: 95450119-95450119 on Build GRCh38
rs1001571103 Chr.5: 95450093-95450094 on Build GRCh38
rs1048721343 Chr.5: 95450094-95450094 on Build GRCh38
rs200730993 Chr.5: 95450280-95450280 on Build GRCh38
rs546333967 Chr.5: 95450137-95450137 on Build GRCh38
rs748322382 Chr.5: 95450268-95450268 on Build GRCh38
rs957538987 Chr.5: 95450255-95450255 on Build GRCh38
rs960108608 Chr.5: 95450196-95450196 on Build GRCh38
rs772577405 Chr.5: 95450163-95450163 on Build GRCh38
rs533320226 Chr.5: 95450307-95450307 on Build GRCh38

View Full Product Details