Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9739833-9740074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF1
Entrez Gene ID
7862
Gene Name
bromodomain and PHD finger containing 1
Gene Aliases
BR140
UniGene
Hs.1004
Gene Chromosome Location
Chr.3: 9731729-9748015 on Build GRCh38
Transcripts
NM_001003694.1 NM_001319049.1 NM_001319050.1 NM_004634.2 XM_005265449.1 XM_005265450.1 XM_005265452.1 XM_011534101.1 XM_011534102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460730 c.1434G>T p.W478C Chr.3: 9739833-9739833 on Build GRCh38
5520851 c.1504G>A p.A502T Chr.3: 9739903-9739903 on Build GRCh38
1049130 c.1472G>A p.R491Q Chr.3: 9739871-9739871 on Build GRCh38
1049129 c.1457C>T p.A486V Chr.3: 9739856-9739856 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967376427 Chr.3: 9740028-9740028 on Build GRCh38
rs369521915 Chr.3: 9739941-9739941 on Build GRCh38
rs755029827 Chr.3: 9739871-9739871 on Build GRCh38
rs140640735 Chr.3: 9739890-9739890 on Build GRCh38
rs778132493 Chr.3: 9739924-9739924 on Build GRCh38
rs931382146 Chr.3: 9740038-9740038 on Build GRCh38
rs769485114 Chr.3: 9740004-9740004 on Build GRCh38
rs547750489 Chr.3: 9739973-9739973 on Build GRCh38
rs756105058 Chr.3: 9739962-9739962 on Build GRCh38
rs749843429 Chr.3: 9739927-9739927 on Build GRCh38
rs1028836336 Chr.3: 9739938-9739938 on Build GRCh38
rs746554710 Chr.3: 9739972-9739972 on Build GRCh38
rs761667095 Chr.3: 9739894-9739898 on Build GRCh38
rs747783020 Chr.3: 9739987-9739987 on Build GRCh38
rs753734128 Chr.3: 9739891-9739892 on Build GRCh38
rs1020242946 Chr.3: 9740017-9740017 on Build GRCh38
rs752949724 Chr.3: 9739910-9739910 on Build GRCh38
rs767673914 Chr.3: 9739892-9739892 on Build GRCh38
rs566167326 Chr.3: 9740026-9740026 on Build GRCh38
rs776207038 Chr.3: 9739983-9739983 on Build GRCh38
rs867377545 Chr.3: 9739915-9739915 on Build GRCh38
rs756445850 Chr.3: 9739913-9739913 on Build GRCh38
rs984307122 Chr.3: 9740040-9740040 on Build GRCh38
rs779536175 Chr.3: 9739968-9739968 on Build GRCh38

View Full Product Details