Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30761023-30761296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC189
Entrez Gene ID
90835
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93
UniGene
Hs.724010
Gene Chromosome Location
Chr.16: 30757226-30762277 on Build GRCh38
Transcripts
NM_001014979.2 NM_001195620.1 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_001014979.2 NM_001195620.1 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148259 c.218G>T p.R73L Chr.16: 30761216-30761216 on Build GRCh38
4151313 c.66C>T p.C22C Chr.16: 30761257-30761257 on Build GRCh38
7148260 c.107G>T p.R36L Chr.16: 30761216-30761216 on Build GRCh38
4151314 c.65G>C p.C22S Chr.16: 30761258-30761258 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913188413 Chr.16: 30761125-30761125 on Build GRCh38
rs768443963 Chr.16: 30761217-30761217 on Build GRCh38
rs765925576 Chr.16: 30761151-30761151 on Build GRCh38
rs758077479 Chr.16: 30761208-30761208 on Build GRCh38
rs774815803 Chr.16: 30761155-30761155 on Build GRCh38
rs764911072 Chr.16: 30761192-30761192 on Build GRCh38
rs985116355 Chr.16: 30761266-30761266 on Build GRCh38
rs765245221 Chr.16: 30761265-30761265 on Build GRCh38
rs933610181 Chr.16: 30761263-30761263 on Build GRCh38
rs758832644 Chr.16: 30761257-30761257 on Build GRCh38
rs534288126 Chr.16: 30761173-30761173 on Build GRCh38
rs544913356 Chr.16: 30761275-30761275 on Build GRCh38
rs750266858 Chr.16: 30761193-30761193 on Build GRCh38
rs143017067 Chr.16: 30761273-30761273 on Build GRCh38
rs753810229 Chr.16: 30761235-30761238 on Build GRCh38
rs373748911 Chr.16: 30761259-30761259 on Build GRCh38
rs749725218 Chr.16: 30761235-30761235 on Build GRCh38
rs528589526 Chr.16: 30761142-30761142 on Build GRCh38
rs771175523 Chr.16: 30761152-30761152 on Build GRCh38
rs886051922 Chr.16: 30761085-30761085 on Build GRCh38
rs753375779 Chr.16: 30761201-30761201 on Build GRCh38
rs1038123542 Chr.16: 30761147-30761147 on Build GRCh38
rs369677903 Chr.16: 30761254-30761254 on Build GRCh38
rs369131280 Chr.16: 30761115-30761115 on Build GRCh38
rs753706982 Chr.16: 30761184-30761184 on Build GRCh38
rs775686090 Chr.16: 30761248-30761248 on Build GRCh38
rs138070048 Chr.16: 30761234-30761234 on Build GRCh38
rs770371321 Chr.16: 30761148-30761148 on Build GRCh38
rs777473706 Chr.16: 30761228-30761236 on Build GRCh38
rs969944689 Chr.16: 30761133-30761133 on Build GRCh38
rs868375268 Chr.16: 30761141-30761141 on Build GRCh38
rs1043628694 Chr.16: 30761277-30761277 on Build GRCh38
rs748995449 Chr.16: 30761078-30761078 on Build GRCh38
rs554975918 Chr.16: 30761045-30761045 on Build GRCh38
rs752321801 Chr.16: 30761191-30761194 on Build GRCh38
rs925252976 Chr.16: 30761081-30761081 on Build GRCh38
rs773087052 Chr.16: 30761272-30761272 on Build GRCh38
rs1045617413 Chr.16: 30761138-30761138 on Build GRCh38
rs754518508 Chr.16: 30761233-30761233 on Build GRCh38
rs927052530 Chr.16: 30761139-30761139 on Build GRCh38
rs779869821 Chr.16: 30761219-30761219 on Build GRCh38
rs746243196 Chr.16: 30761245-30761245 on Build GRCh38
rs367805411 Chr.16: 30761145-30761145 on Build GRCh38
rs149467590 Chr.16: 30761244-30761244 on Build GRCh38
rs751051814 Chr.16: 30761223-30761223 on Build GRCh38
rs1032108501 Chr.16: 30761114-30761114 on Build GRCh38
rs778134113 Chr.16: 30761149-30761149 on Build GRCh38
rs1022868307 Chr.16: 30761128-30761128 on Build GRCh38
rs761860197 Chr.16: 30761191-30761191 on Build GRCh38
rs764301654 Chr.16: 30761182-30761182 on Build GRCh38
rs573325591 Chr.16: 30761104-30761104 on Build GRCh38
rs772301654 Chr.16: 30761171-30761171 on Build GRCh38
rs760747798 Chr.16: 30761174-30761174 on Build GRCh38
rs964504253 Chr.16: 30761074-30761074 on Build GRCh38
rs1014131805 Chr.16: 30761119-30761119 on Build GRCh38
rs1018340647 Chr.16: 30761055-30761055 on Build GRCh38
rs759713220 Chr.16: 30761160-30761160 on Build GRCh38
rs984382146 Chr.16: 30761159-30761159 on Build GRCh38
rs761619254 Chr.16: 30761264-30761264 on Build GRCh38
rs771497269 Chr.16: 30761243-30761243 on Build GRCh38
rs956694807 Chr.16: 30761082-30761082 on Build GRCh38

View Full Product Details