Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26933177-26933414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL5
Entrez Gene ID
56896
Gene Name
dihydropyrimidinase like 5
Gene Aliases
CRAM, CRMP-5, CRMP5, Ulip6
UniGene
Hs.299315
Gene Chromosome Location
Chr.2: 26848101-26950351 on Build GRCh38
Transcripts
NM_001253723.1 NM_001253724.1 NM_020134.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251069 c.790+1G>T p.? Chr.2: 26933334-26933334 on Build GRCh38
310702 c.763G>T p.D255Y Chr.2: 26933306-26933306 on Build GRCh38
3962718 c.783G>A p.K261K Chr.2: 26933326-26933326 on Build GRCh38
6157678 c.715-1G>T p.? Chr.2: 26933257-26933257 on Build GRCh38
720687 c.755C>T p.S252L Chr.2: 26933298-26933298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918966164 Chr.2: 26933302-26933302 on Build GRCh38
rs748115002 Chr.2: 26933276-26933276 on Build GRCh38
rs149534583 Chr.2: 26933270-26933270 on Build GRCh38
rs943066665 Chr.2: 26933372-26933372 on Build GRCh38
rs749044198 Chr.2: 26933363-26933363 on Build GRCh38
rs772438840 Chr.2: 26933367-26933367 on Build GRCh38
rs780744088 Chr.2: 26933328-26933328 on Build GRCh38
rs143909555 Chr.2: 26933306-26933306 on Build GRCh38
rs757759496 Chr.2: 26933309-26933309 on Build GRCh38
rs141353764 Chr.2: 26933267-26933267 on Build GRCh38
rs765630736 Chr.2: 26933315-26933315 on Build GRCh38
rs754514639 Chr.2: 26933327-26933327 on Build GRCh38
rs768650634 Chr.2: 26933269-26933269 on Build GRCh38
rs779568197 Chr.2: 26933252-26933252 on Build GRCh38
rs773172791 Chr.2: 26933361-26933361 on Build GRCh38
rs989809438 Chr.2: 26933305-26933305 on Build GRCh38
rs757628029 Chr.2: 26933219-26933219 on Build GRCh38
rs767809586 Chr.2: 26933295-26933295 on Build GRCh38
rs888792727 Chr.2: 26933360-26933360 on Build GRCh38
rs754231655 Chr.2: 26933217-26933217 on Build GRCh38
rs745568928 Chr.2: 26933234-26933234 on Build GRCh38
rs184784357 Chr.2: 26933232-26933232 on Build GRCh38
rs760937177 Chr.2: 26933375-26933375 on Build GRCh38
rs538138952 Chr.2: 26933304-26933304 on Build GRCh38
rs758011185 Chr.2: 26933235-26933235 on Build GRCh38
rs779621495 Chr.2: 26933322-26933322 on Build GRCh38
rs746529100 Chr.2: 26933326-26933326 on Build GRCh38
rs754358873 Chr.2: 26933308-26933308 on Build GRCh38
rs769538594 Chr.2: 26933284-26933284 on Build GRCh38
rs769739477 Chr.2: 26933359-26933359 on Build GRCh38
rs775918938 Chr.2: 26933371-26933371 on Build GRCh38
rs768942257 Chr.2: 26933220-26933220 on Build GRCh38
rs773123619 Chr.2: 26933287-26933287 on Build GRCh38
rs949906335 Chr.2: 26933290-26933290 on Build GRCh38
rs759878048 Chr.2: 26933294-26933294 on Build GRCh38
rs748089233 Chr.2: 26933342-26933342 on Build GRCh38
rs1009783486 Chr.2: 26933351-26933351 on Build GRCh38
rs1040584959 Chr.2: 26933389-26933389 on Build GRCh38
rs189492736 Chr.2: 26933299-26933299 on Build GRCh38
rs758066334 Chr.2: 26933319-26933319 on Build GRCh38
rs372829541 Chr.2: 26933298-26933298 on Build GRCh38
rs750743114 Chr.2: 26933316-26933316 on Build GRCh38

View Full Product Details