Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49066155-49066424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSE1L
Entrez Gene ID
1434
Gene Name
chromosome segregation 1 like
Gene Aliases
CAS, CSE1, XPO2
UniGene
Hs.90073
Gene Chromosome Location
Chr.20: 49046246-49096960 on Build GRCh38
Transcripts
NM_001256135.1 NM_001316.3 NR_045796.1 XM_011528599.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362782 c.385G>C p.D129H Chr.20: 49066419-49066419 on Build GRCh38
5439731 c.284A>G p.N95S Chr.20: 49066247-49066247 on Build GRCh38
6308378 c.280G>A p.A94T Chr.20: 49066243-49066243 on Build GRCh38
7105507 c.261C>T p.A87A Chr.20: 49066224-49066224 on Build GRCh38
189822 c.265C>T p.R89* Chr.20: 49066228-49066228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370077115 Chr.20: 49066400-49066400 on Build GRCh38
rs771895995 Chr.20: 49066299-49066299 on Build GRCh38
rs777393666 Chr.20: 49066358-49066358 on Build GRCh38
rs562112551 Chr.20: 49066236-49066236 on Build GRCh38
rs141564653 Chr.20: 49066203-49066203 on Build GRCh38
rs993625726 Chr.20: 49066277-49066277 on Build GRCh38
rs745440251 Chr.20: 49066383-49066383 on Build GRCh38
rs751766380 Chr.20: 49066335-49066335 on Build GRCh38
rs770505854 Chr.20: 49066275-49066275 on Build GRCh38
rs181128748 Chr.20: 49066323-49066323 on Build GRCh38
rs762589431 Chr.20: 49066204-49066204 on Build GRCh38
rs1024466852 Chr.20: 49066246-49066246 on Build GRCh38
rs2246266 Chr.20: 49066356-49066356 on Build GRCh38
rs780226225 Chr.20: 49066247-49066247 on Build GRCh38
rs201843660 Chr.20: 49066207-49066207 on Build GRCh38
rs765377854 Chr.20: 49066322-49066322 on Build GRCh38
rs376007852 Chr.20: 49066319-49066319 on Build GRCh38
rs771697950 Chr.20: 49066387-49066387 on Build GRCh38
rs768020549 Chr.20: 49066217-49066217 on Build GRCh38
rs775291464 Chr.20: 49066391-49066391 on Build GRCh38
rs150897794 Chr.20: 49066302-49066302 on Build GRCh38
rs898000207 Chr.20: 49066258-49066258 on Build GRCh38
rs566431913 Chr.20: 49066307-49066307 on Build GRCh38
rs778882666 Chr.20: 49066377-49066377 on Build GRCh38
rs754283301 Chr.20: 49066229-49066229 on Build GRCh38
rs770826162 Chr.20: 49066368-49066368 on Build GRCh38
rs773686690 Chr.20: 49066276-49066276 on Build GRCh38
rs749114451 Chr.20: 49066268-49066268 on Build GRCh38
rs764767332 Chr.20: 49066228-49066228 on Build GRCh38
rs377396767 Chr.20: 49066244-49066244 on Build GRCh38
rs775732416 Chr.20: 49066347-49066350 on Build GRCh38
rs759072407 Chr.20: 49066210-49066210 on Build GRCh38
rs111799496 Chr.20: 49066346-49066346 on Build GRCh38
rs953962923 Chr.20: 49066325-49066325 on Build GRCh38
rs780319629 Chr.20: 49066326-49066326 on Build GRCh38
rs753165495 Chr.20: 49066221-49066221 on Build GRCh38
rs200581090 Chr.20: 49066193-49066193 on Build GRCh38
rs879301645 Chr.20: 49066344-49066344 on Build GRCh38
rs756518336 Chr.20: 49066224-49066224 on Build GRCh38
rs749270353 Chr.20: 49066365-49066365 on Build GRCh38
rs774984847 Chr.20: 49066373-49066373 on Build GRCh38
rs759068057 Chr.20: 49066288-49066288 on Build GRCh38
rs778787944 Chr.20: 49066237-49066237 on Build GRCh38
rs1027925485 Chr.20: 49066278-49066278 on Build GRCh38
rs769571209 Chr.20: 49066250-49066250 on Build GRCh38
rs758565893 Chr.20: 49066245-49066245 on Build GRCh38
rs113366638 Chr.20: 49066301-49066301 on Build GRCh38
rs529763386 Chr.20: 49066248-49066248 on Build GRCh38
rs777781671 Chr.20: 49066256-49066256 on Build GRCh38

View Full Product Details