Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96797446-96797697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
UniGene
Hs.175043
Gene Chromosome Location
Chr.2: 96760902-96811891 on Build GRCh38
Transcripts
NM_020184.3 XM_005263914.3 XM_005263915.3 XM_011510955.2 XM_017003799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841271 c.28C>T p.R10W Chr.2: 96797533-96797533 on Build GRCh38
1284184 c.1551C>A p.D517E Chr.2: 96797517-96797517 on Build GRCh38
5528827 c.1671C>T p.F557F Chr.2: 96797637-96797637 on Build GRCh38
4316866 c.105G>A p.P35P Chr.2: 96797610-96797610 on Build GRCh38
5603232 c.1642C>T p.P548S Chr.2: 96797608-96797608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950027489 Chr.2: 96797539-96797539 on Build GRCh38
rs747798390 Chr.2: 96797678-96797678 on Build GRCh38
rs775648616 Chr.2: 96797554-96797554 on Build GRCh38
rs1019151584 Chr.2: 96797676-96797676 on Build GRCh38
rs766428142 Chr.2: 96797655-96797655 on Build GRCh38
rs748985173 Chr.2: 96797472-96797472 on Build GRCh38
rs1055800062 Chr.2: 96797596-96797596 on Build GRCh38
rs761645419 Chr.2: 96797627-96797627 on Build GRCh38
rs763026644 Chr.2: 96797625-96797625 on Build GRCh38
rs1052978770 Chr.2: 96797517-96797517 on Build GRCh38
rs764314767 Chr.2: 96797522-96797522 on Build GRCh38
rs145616864 Chr.2: 96797497-96797497 on Build GRCh38
rs773659426 Chr.2: 96797482-96797482 on Build GRCh38
rs753328499 Chr.2: 96797668-96797668 on Build GRCh38
rs942844035 Chr.2: 96797476-96797476 on Build GRCh38
rs756063575 Chr.2: 96797654-96797654 on Build GRCh38
rs201332826 Chr.2: 96797605-96797605 on Build GRCh38
rs771253370 Chr.2: 96797579-96797579 on Build GRCh38
rs755653851 Chr.2: 96797549-96797549 on Build GRCh38
rs371651491 Chr.2: 96797496-96797496 on Build GRCh38
rs769244069 Chr.2: 96797470-96797470 on Build GRCh38
rs374104702 Chr.2: 96797638-96797638 on Build GRCh38
rs34834618 Chr.2: 96797609-96797610 on Build GRCh38
rs368275804 Chr.2: 96797507-96797507 on Build GRCh38
rs761236064 Chr.2: 96797609-96797609 on Build GRCh38
rs770361608 Chr.2: 96797637-96797637 on Build GRCh38
rs367780275 Chr.2: 96797488-96797488 on Build GRCh38
rs781294642 Chr.2: 96797651-96797651 on Build GRCh38
rs779829101 Chr.2: 96797471-96797471 on Build GRCh38
rs1002208092 Chr.2: 96797598-96797598 on Build GRCh38
rs772498225 Chr.2: 96797527-96797527 on Build GRCh38
rs767254813 Chr.2: 96797632-96797632 on Build GRCh38
rs762522792 Chr.2: 96797543-96797545 on Build GRCh38
rs753964734 Chr.2: 96797673-96797673 on Build GRCh38
rs752807120 Chr.2: 96797519-96797519 on Build GRCh38
rs764294792 Chr.2: 96797626-96797626 on Build GRCh38
rs764568597 Chr.2: 96797610-96797610 on Build GRCh38
rs746322998 Chr.2: 96797587-96797587 on Build GRCh38
rs745378599 Chr.2: 96797469-96797469 on Build GRCh38
rs750374054 Chr.2: 96797534-96797534 on Build GRCh38
rs1006079657 Chr.2: 96797576-96797576 on Build GRCh38
rs569907931 Chr.2: 96797553-96797553 on Build GRCh38
rs145764649 Chr.2: 96797533-96797533 on Build GRCh38
rs148365594 Chr.2: 96797528-96797528 on Build GRCh38
rs762463681 Chr.2: 96797508-96797508 on Build GRCh38
rs1045671384 Chr.2: 96797544-96797544 on Build GRCh38
rs768361992 Chr.2: 96797555-96797555 on Build GRCh38
rs971025370 Chr.2: 96797652-96797652 on Build GRCh38
rs747389796 Chr.2: 96797578-96797578 on Build GRCh38
rs554605541 Chr.2: 96797669-96797669 on Build GRCh38
rs763115064 Chr.2: 96797520-96797520 on Build GRCh38
rs777167557 Chr.2: 96797580-96797580 on Build GRCh38

View Full Product Details