Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14720209-14720458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CER1
Entrez Gene ID
9350
Gene Name
cerberus 1, DAN family BMP antagonist
Gene Aliases
DAND4
UniGene
Hs.248204
Gene Chromosome Location
Chr.9: 14715082-14724308 on Build GRCh38
Transcripts
NM_005454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3656170 c.586C>T p.P196S Chr.9: 14720308-14720308 on Build GRCh38
1235110 c.578T>A p.V193D Chr.9: 14720316-14720316 on Build GRCh38
4671758 c.571G>A p.G191R Chr.9: 14720323-14720323 on Build GRCh38
6489758 c.648G>A p.T216T Chr.9: 14720246-14720246 on Build GRCh38
1248113 c.638A>C p.K213T Chr.9: 14720256-14720256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764877021 Chr.9: 14720422-14720422 on Build GRCh38
rs369153749 Chr.9: 14720349-14720349 on Build GRCh38
rs375135027 Chr.9: 14720263-14720263 on Build GRCh38
rs201357342 Chr.9: 14720374-14720374 on Build GRCh38
rs148470660 Chr.9: 14720240-14720240 on Build GRCh38
rs140434620 Chr.9: 14720298-14720298 on Build GRCh38
rs527955982 Chr.9: 14720297-14720297 on Build GRCh38
rs112686852 Chr.9: 14720370-14720370 on Build GRCh38
rs780114954 Chr.9: 14720288-14720288 on Build GRCh38
rs765944833 Chr.9: 14720247-14720247 on Build GRCh38
rs760306709 Chr.9: 14720246-14720246 on Build GRCh38
rs147692802 Chr.9: 14720342-14720342 on Build GRCh38
rs35706018 Chr.9: 14720360-14720360 on Build GRCh38
rs145380330 Chr.9: 14720306-14720306 on Build GRCh38
rs371412418 Chr.9: 14720413-14720413 on Build GRCh38
rs142706502 Chr.9: 14720244-14720244 on Build GRCh38
rs377725836 Chr.9: 14720255-14720255 on Build GRCh38
rs747007452 Chr.9: 14720399-14720399 on Build GRCh38
rs775193222 Chr.9: 14720430-14720430 on Build GRCh38
rs752024589 Chr.9: 14720260-14720260 on Build GRCh38
rs749914334 Chr.9: 14720245-14720245 on Build GRCh38
rs774395535 Chr.9: 14720241-14720241 on Build GRCh38
rs201012414 Chr.9: 14720259-14720260 on Build GRCh38
rs748038785 Chr.9: 14720299-14720299 on Build GRCh38
rs773003343 Chr.9: 14720392-14720392 on Build GRCh38
rs533058950 Chr.9: 14720324-14720324 on Build GRCh38
rs111351331 Chr.9: 14720412-14720412 on Build GRCh38
rs772051591 Chr.9: 14720383-14720383 on Build GRCh38
rs372214721 Chr.9: 14720348-14720348 on Build GRCh38
rs755263838 Chr.9: 14720365-14720365 on Build GRCh38
rs17289263 Chr.9: 14720363-14720363 on Build GRCh38
rs1006445515 Chr.9: 14720341-14720341 on Build GRCh38
rs759221042 Chr.9: 14720323-14720323 on Build GRCh38
rs756273156 Chr.9: 14720286-14720286 on Build GRCh38
rs755576353 Chr.9: 14720396-14720396 on Build GRCh38
rs749561433 Chr.9: 14720292-14720292 on Build GRCh38
rs367962269 Chr.9: 14720233-14720233 on Build GRCh38
rs7036635 Chr.9: 14720359-14720359 on Build GRCh38
rs781192924 Chr.9: 14720279-14720279 on Build GRCh38
rs369930781 Chr.9: 14720301-14720301 on Build GRCh38
rs533770342 Chr.9: 14720235-14720235 on Build GRCh38
rs146765149 Chr.9: 14720249-14720249 on Build GRCh38
rs995927573 Chr.9: 14720229-14720229 on Build GRCh38
rs770307177 Chr.9: 14720407-14720407 on Build GRCh38
rs753586509 Chr.9: 14720248-14720248 on Build GRCh38
rs202189907 Chr.9: 14720316-14720316 on Build GRCh38
rs772139341 Chr.9: 14720300-14720300 on Build GRCh38
rs745943371 Chr.9: 14720280-14720280 on Build GRCh38
rs761873382 Chr.9: 14720242-14720242 on Build GRCh38
rs970445894 Chr.9: 14720385-14720386 on Build GRCh38
rs765963499 Chr.9: 14720310-14720310 on Build GRCh38
rs999315446 Chr.9: 14720393-14720393 on Build GRCh38
rs1053227464 Chr.9: 14720330-14720330 on Build GRCh38
rs138899880 Chr.9: 14720375-14720375 on Build GRCh38

View Full Product Details