Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18868647-18868896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS1
Entrez Gene ID
10715
Gene Name
ceramide synthase 1
Gene Aliases
EPM8, LAG1, LASS1, UOG1
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896727 on Build GRCh38
Transcripts
NM_021267.4 NM_001492.5

Gene Symbol
GDF1
Entrez Gene ID
2657
Gene Name
growth differentiation factor 1
Gene Aliases
DORV, DTGA3, RAI
UniGene
Hs.412355
Gene Chromosome Location
Chr.19: 18868546-18896144 on Build GRCh38
Transcripts
NM_021267.4 NM_001492.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391748 c.1012G>A p.V338M Chr.19: 18868704-18868704 on Build GRCh38
1208048 c.914C>T p.A305V Chr.19: 18868802-18868802 on Build GRCh38
6585868 c.865C>T p.R289C Chr.19: 18868851-18868851 on Build GRCh38
6457604 c.921G>A p.S307S Chr.19: 18868795-18868795 on Build GRCh38
5626742 c.912C>T p.V304V Chr.19: 18868804-18868804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764119124 Chr.19: 18868796-18868796 on Build GRCh38
rs769364770 Chr.19: 18868674-18868674 on Build GRCh38
rs551588378 Chr.19: 18868804-18868804 on Build GRCh38
rs753906369 Chr.19: 18868757-18868757 on Build GRCh38
rs1008969968 Chr.19: 18868840-18868840 on Build GRCh38
rs773271648 Chr.19: 18868729-18868729 on Build GRCh38
rs1057491518 Chr.19: 18868722-18868722 on Build GRCh38
rs760908706 Chr.19: 18868731-18868731 on Build GRCh38
rs762069368 Chr.19: 18868702-18868702 on Build GRCh38
rs748808313 Chr.19: 18868687-18868687 on Build GRCh38
rs756972470 Chr.19: 18868856-18868856 on Build GRCh38
rs945025237 Chr.19: 18868681-18868681 on Build GRCh38
rs766727386 Chr.19: 18868732-18868732 on Build GRCh38
rs864622513 Chr.19: 18868791-18868791 on Build GRCh38
rs948868401 Chr.19: 18868765-18868765 on Build GRCh38
rs768027510 Chr.19: 18868666-18868669 on Build GRCh38
rs1037920678 Chr.19: 18868769-18868769 on Build GRCh38
rs606231383 Chr.19: 18868806-18868807 on Build GRCh38
rs769083453 Chr.19: 18868692-18868692 on Build GRCh38
rs779492558 Chr.19: 18868686-18868686 on Build GRCh38
rs537524649 Chr.19: 18868697-18868697 on Build GRCh38
rs772532758 Chr.19: 18868711-18868711 on Build GRCh38
rs890558836 Chr.19: 18868825-18868825 on Build GRCh38
rs1026312296 Chr.19: 18868816-18868816 on Build GRCh38
rs894657723 Chr.19: 18868740-18868740 on Build GRCh38
rs759759832 Chr.19: 18868781-18868781 on Build GRCh38
rs376692379 Chr.19: 18868717-18868717 on Build GRCh38
rs865998453 Chr.19: 18868701-18868701 on Build GRCh38
rs867919965 Chr.19: 18868848-18868848 on Build GRCh38
rs899391416 Chr.19: 18868788-18868788 on Build GRCh38

View Full Product Details