Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 170322856-170323129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM120B
Entrez Gene ID
84498
Gene Name
family with sequence similarity 120B
Gene Aliases
CCPG, KIAA1838, PGCC1, dJ894D12.1
UniGene
Hs.369522
Gene Chromosome Location
Chr.6: 170290703-170407748 on Build GRCh38
Transcripts
NM_001286379.1 NM_001286380.1 NM_001286381.1 NM_032448.2 XM_011536185.2 XM_017011357.1 XM_017011358.1 XM_017011359.1 XM_017011360.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3860560 c.1753G>A p.V585I Chr.6: 170323097-170323097 on Build GRCh38
7146448 c.1781A>G p.N594S Chr.6: 170323125-170323125 on Build GRCh38
365869 c.1776G>C p.V592V Chr.6: 170323120-170323120 on Build GRCh38
7331929 c.1744A>G p.T582A Chr.6: 170323088-170323088 on Build GRCh38
6658231 c.1752C>T p.H584H Chr.6: 170323096-170323096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370272691 Chr.6: 170323103-170323103 on Build GRCh38
rs375963765 Chr.6: 170323076-170323076 on Build GRCh38
rs773198525 Chr.6: 170323080-170323080 on Build GRCh38
rs944467260 Chr.6: 170323017-170323017 on Build GRCh38
rs769944678 Chr.6: 170323064-170323064 on Build GRCh38
rs759194732 Chr.6: 170323032-170323032 on Build GRCh38
rs1036004329 Chr.6: 170323026-170323026 on Build GRCh38
rs745895342 Chr.6: 170323044-170323044 on Build GRCh38
rs533306663 Chr.6: 170322925-170322925 on Build GRCh38
rs1038348696 Chr.6: 170322917-170322917 on Build GRCh38
rs144634371 Chr.6: 170323096-170323096 on Build GRCh38
rs548504885 Chr.6: 170322966-170322966 on Build GRCh38
rs890997468 Chr.6: 170323021-170323021 on Build GRCh38
rs570549179 Chr.6: 170323073-170323073 on Build GRCh38
rs940019468 Chr.6: 170322893-170322893 on Build GRCh38
rs2064532 Chr.6: 170322976-170322976 on Build GRCh38
rs552822243 Chr.6: 170323097-170323097 on Build GRCh38
rs781358299 Chr.6: 170322880-170322880 on Build GRCh38
rs140670595 Chr.6: 170323079-170323079 on Build GRCh38
rs560109864 Chr.6: 170322943-170322943 on Build GRCh38
rs530952725 Chr.6: 170323031-170323031 on Build GRCh38
rs868121837 Chr.6: 170323045-170323045 on Build GRCh38
rs749201013 Chr.6: 170322929-170322929 on Build GRCh38
rs187776926 Chr.6: 170322956-170322956 on Build GRCh38
rs78865601 Chr.6: 170323033-170323033 on Build GRCh38
rs781399279 Chr.6: 170323035-170323035 on Build GRCh38

View Full Product Details