Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128325704-128325959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ4
Entrez Gene ID
51117
Gene Name
coenzyme Q4
Gene Aliases
CGI-92, COQ10D7
UniGene
Hs.98541
Gene Chromosome Location
Chr.9: 128322486-128334072 on Build GRCh38
Transcripts
NM_001305942.1 NM_016035.4 XM_017014792.1 XM_017014793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782278 c.385C>G p.R129G Chr.9: 128325864-128325864 on Build GRCh38
5977093 c.386G>A p.R129H Chr.9: 128325865-128325865 on Build GRCh38
1284264 c.339C>A p.G113G Chr.9: 128325818-128325818 on Build GRCh38
5431617 c.329T>G p.L110R Chr.9: 128325808-128325808 on Build GRCh38
5947016 c.393G>C p.L131L Chr.9: 128325872-128325872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761787873 Chr.9: 128325787-128325787 on Build GRCh38
rs773038253 Chr.9: 128325841-128325841 on Build GRCh38
rs767379261 Chr.9: 128325796-128325796 on Build GRCh38
rs748735656 Chr.9: 128325744-128325744 on Build GRCh38
rs779410332 Chr.9: 128325740-128325740 on Build GRCh38
rs1046811704 Chr.9: 128325789-128325789 on Build GRCh38
rs748977925 Chr.9: 128325818-128325818 on Build GRCh38
rs770974172 Chr.9: 128325922-128325922 on Build GRCh38
rs778289915 Chr.9: 128325751-128325751 on Build GRCh38
rs768504691 Chr.9: 128325782-128325782 on Build GRCh38
rs77448374 Chr.9: 128325728-128325728 on Build GRCh38
rs572683310 Chr.9: 128325790-128325790 on Build GRCh38
rs541118265 Chr.9: 128325771-128325771 on Build GRCh38
rs958071474 Chr.9: 128325932-128325932 on Build GRCh38
rs375096994 Chr.9: 128325854-128325854 on Build GRCh38
rs776664599 Chr.9: 128325929-128325929 on Build GRCh38
rs558324741 Chr.9: 128325886-128325886 on Build GRCh38
rs534077160 Chr.9: 128325826-128325826 on Build GRCh38
rs747779231 Chr.9: 128325882-128325882 on Build GRCh38
rs759199443 Chr.9: 128325751-128325752 on Build GRCh38
rs769933028 Chr.9: 128325938-128325938 on Build GRCh38
rs779533723 Chr.9: 128325815-128325815 on Build GRCh38
rs754124683 Chr.9: 128325918-128325918 on Build GRCh38
rs369623109 Chr.9: 128325727-128325727 on Build GRCh38
rs929713295 Chr.9: 128325784-128325784 on Build GRCh38
rs752070573 Chr.9: 128325857-128325857 on Build GRCh38
rs758803466 Chr.9: 128325749-128325749 on Build GRCh38
rs745396359 Chr.9: 128325754-128325754 on Build GRCh38
rs776825296 Chr.9: 128325849-128325849 on Build GRCh38
rs368669078 Chr.9: 128325902-128325902 on Build GRCh38
rs754174993 Chr.9: 128325800-128325800 on Build GRCh38
rs760650266 Chr.9: 128325846-128325846 on Build GRCh38
rs753368044 Chr.9: 128325870-128325870 on Build GRCh38
rs753487079 Chr.9: 128325743-128325743 on Build GRCh38
rs371984550 Chr.9: 128325783-128325783 on Build GRCh38
rs765741653 Chr.9: 128325803-128325803 on Build GRCh38
rs755249411 Chr.9: 128325802-128325802 on Build GRCh38
rs754048442 Chr.9: 128325737-128325737 on Build GRCh38
rs752897625 Chr.9: 128325853-128325853 on Build GRCh38
rs976601893 Chr.9: 128325768-128325768 on Build GRCh38
rs768254447 Chr.9: 128325747-128325747 on Build GRCh38
rs758745776 Chr.9: 128325809-128325809 on Build GRCh38
rs571683970 Chr.9: 128325735-128325735 on Build GRCh38
rs111535641 Chr.9: 128325863-128325863 on Build GRCh38
rs753093771 Chr.9: 128325805-128325805 on Build GRCh38
rs138917651 Chr.9: 128325836-128325836 on Build GRCh38
rs141303335 Chr.9: 128325777-128325777 on Build GRCh38
rs773943371 Chr.9: 128325835-128325835 on Build GRCh38
rs770985856 Chr.9: 128325848-128325848 on Build GRCh38
rs748924643 Chr.9: 128325780-128325780 on Build GRCh38
rs2270203 Chr.9: 128325797-128325797 on Build GRCh38
rs1025159224 Chr.9: 128325827-128325827 on Build GRCh38
rs778542323 Chr.9: 128325881-128325881 on Build GRCh38
rs764762017 Chr.9: 128325755-128325756 on Build GRCh38
rs766426220 Chr.9: 128325798-128325798 on Build GRCh38
rs372569872 Chr.9: 128325738-128325738 on Build GRCh38
rs149398860 Chr.9: 128325855-128325855 on Build GRCh38
rs942554091 Chr.9: 128325763-128325763 on Build GRCh38
rs777622469 Chr.9: 128325894-128325894 on Build GRCh38
rs530213004 Chr.9: 128325810-128325810 on Build GRCh38
rs199661029 Chr.9: 128325852-128325852 on Build GRCh38
rs139770523 Chr.9: 128325864-128325864 on Build GRCh38
rs754445455 Chr.9: 128325873-128325873 on Build GRCh38
rs79172715 Chr.9: 128325935-128325935 on Build GRCh38

View Full Product Details