Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 63960811-63961082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR1A
Entrez Gene ID
3350
Gene Name
5-hydroxytryptamine receptor 1A
Gene Aliases
5-HT-1A, 5-HT1A, 5HT1a, ADRB2RL1, ADRBRL1, G-21, PFMCD
UniGene
Hs.247940
Gene Chromosome Location
Chr.5: 63960048-63962292 on Build GRCh38
Transcripts
NM_000524.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1743531 c.782G>T p.R261M Chr.5: 63960938-63960938 on Build GRCh38
6171663 c.643T>C p.Y215H Chr.5: 63961077-63961077 on Build GRCh38
4756442 c.752A>C p.K251T Chr.5: 63960968-63960968 on Build GRCh38
3617356 c.753G>A p.K251K Chr.5: 63960967-63960967 on Build GRCh38
6682874 c.738C>T p.P246P Chr.5: 63960982-63960982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771552839 Chr.5: 63960906-63960906 on Build GRCh38
rs749883886 Chr.5: 63960946-63960946 on Build GRCh38
rs1800042 Chr.5: 63960902-63960902 on Build GRCh38
rs754879239 Chr.5: 63961009-63961009 on Build GRCh38
rs138074971 Chr.5: 63960903-63960903 on Build GRCh38
rs539402086 Chr.5: 63960955-63960955 on Build GRCh38
rs777222317 Chr.5: 63960913-63960913 on Build GRCh38
rs761266205 Chr.5: 63960947-63960947 on Build GRCh38
rs201179587 Chr.5: 63961004-63961004 on Build GRCh38
rs200711760 Chr.5: 63961003-63961003 on Build GRCh38
rs1029046608 Chr.5: 63960971-63960971 on Build GRCh38
rs555710952 Chr.5: 63961053-63961053 on Build GRCh38
rs755997333 Chr.5: 63960868-63960868 on Build GRCh38
rs372264021 Chr.5: 63960897-63960897 on Build GRCh38
rs753733145 Chr.5: 63960967-63960967 on Build GRCh38
rs200441950 Chr.5: 63960847-63960847 on Build GRCh38
rs779879578 Chr.5: 63960877-63960877 on Build GRCh38
rs149284354 Chr.5: 63960865-63960865 on Build GRCh38
rs75236124 Chr.5: 63961002-63961002 on Build GRCh38
rs865852563 Chr.5: 63960945-63960945 on Build GRCh38
rs200926929 Chr.5: 63960880-63960880 on Build GRCh38
rs758546216 Chr.5: 63960980-63960980 on Build GRCh38
rs536132374 Chr.5: 63961006-63961006 on Build GRCh38
rs775709782 Chr.5: 63960998-63960998 on Build GRCh38
rs761956170 Chr.5: 63960831-63960831 on Build GRCh38
rs34158987 Chr.5: 63960921-63960921 on Build GRCh38
rs763855576 Chr.5: 63960859-63960859 on Build GRCh38
rs771070406 Chr.5: 63960986-63960986 on Build GRCh38
rs755560498 Chr.5: 63960949-63960949 on Build GRCh38
rs925000687 Chr.5: 63960834-63960834 on Build GRCh38
rs896078356 Chr.5: 63960983-63960983 on Build GRCh38
rs767087151 Chr.5: 63961000-63961000 on Build GRCh38
rs1800044 Chr.5: 63961061-63961061 on Build GRCh38
rs758155850 Chr.5: 63961013-63961013 on Build GRCh38
rs367863011 Chr.5: 63961042-63961042 on Build GRCh38
rs761581962 Chr.5: 63960999-63960999 on Build GRCh38
rs770336424 Chr.5: 63960994-63960994 on Build GRCh38
rs761353761 Chr.5: 63960935-63960935 on Build GRCh38
rs757438767 Chr.5: 63961052-63961052 on Build GRCh38
rs202078108 Chr.5: 63960981-63960981 on Build GRCh38
rs748443642 Chr.5: 63960976-63960976 on Build GRCh38
rs759986249 Chr.5: 63960914-63960914 on Build GRCh38
rs142264665 Chr.5: 63960990-63960990 on Build GRCh38
rs150835535 Chr.5: 63960992-63960992 on Build GRCh38
rs747075229 Chr.5: 63960982-63960982 on Build GRCh38
rs772003513 Chr.5: 63960837-63960837 on Build GRCh38
rs200472121 Chr.5: 63960904-63960904 on Build GRCh38
rs777864947 Chr.5: 63961024-63961024 on Build GRCh38
rs148698436 Chr.5: 63960972-63960972 on Build GRCh38
rs143647030 Chr.5: 63960919-63960919 on Build GRCh38
rs373988348 Chr.5: 63960899-63960899 on Build GRCh38
rs138438693 Chr.5: 63960856-63960856 on Build GRCh38
rs761065498 Chr.5: 63960840-63960840 on Build GRCh38
rs746421729 Chr.5: 63961055-63961055 on Build GRCh38
rs201899093 Chr.5: 63961011-63961011 on Build GRCh38
rs199801230 Chr.5: 63961012-63961012 on Build GRCh38
rs372438845 Chr.5: 63961057-63961057 on Build GRCh38
rs199635945 Chr.5: 63960862-63960862 on Build GRCh38
rs957870350 Chr.5: 63960849-63960849 on Build GRCh38
rs754252782 Chr.5: 63960853-63960853 on Build GRCh38
rs879251256 Chr.5: 63961034-63961034 on Build GRCh38
rs201463384 Chr.5: 63960883-63960883 on Build GRCh38
rs202118060 Chr.5: 63960838-63960838 on Build GRCh38
rs999108709 Chr.5: 63961033-63961033 on Build GRCh38
rs767117374 Chr.5: 63960943-63960943 on Build GRCh38
rs147211774 Chr.5: 63960962-63960962 on Build GRCh38

View Full Product Details