Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107416168-107416397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWF19L2
Entrez Gene ID
143884
Gene Name
CWF19-like 2, cell cycle control (S. pombe)
Gene Aliases
-
UniGene
Hs.212140
Gene Chromosome Location
Chr.11: 107326345-107457846 on Build GRCh38
Transcripts
NM_152434.2 XM_017017239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3666064 c.1154A>G p.E385G Chr.11: 107416210-107416210 on Build GRCh38
5452258 c.1609G>A p.G537R Chr.11: 107416217-107416217 on Build GRCh38
922588 c.1139A>C p.K380T Chr.11: 107416225-107416225 on Build GRCh38
5452257 c.1147G>A p.G383R Chr.11: 107416217-107416217 on Build GRCh38
4795451 c.1148G>A p.G383E Chr.11: 107416216-107416216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896656535 Chr.11: 107416350-107416352 on Build GRCh38
rs895758137 Chr.11: 107416371-107416371 on Build GRCh38
rs747613461 Chr.11: 107416330-107416330 on Build GRCh38
rs539047046 Chr.11: 107416351-107416351 on Build GRCh38
rs905682357 Chr.11: 107416338-107416338 on Build GRCh38
rs758878293 Chr.11: 107416307-107416307 on Build GRCh38
rs931023806 Chr.11: 107416309-107416309 on Build GRCh38
rs1002716265 Chr.11: 107416231-107416231 on Build GRCh38
rs572841314 Chr.11: 107416248-107416248 on Build GRCh38
rs764310573 Chr.11: 107416372-107416372 on Build GRCh38
rs17106909 Chr.11: 107416217-107416217 on Build GRCh38
rs918340686 Chr.11: 107416219-107416219 on Build GRCh38
rs898601986 Chr.11: 107416222-107416222 on Build GRCh38
rs769602879 Chr.11: 107416332-107416332 on Build GRCh38
rs369574894 Chr.11: 107416352-107416352 on Build GRCh38
rs868105735 Chr.11: 107416221-107416221 on Build GRCh38
rs185913800 Chr.11: 107416310-107416310 on Build GRCh38

View Full Product Details