Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 13092487-13092752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLC1
Entrez Gene ID
10395
Gene Name
DLC1 Rho GTPase activating protein
Gene Aliases
ARHGAP7, HP, STARD12, p122-RhoGAP
UniGene
Hs.134296
Gene Chromosome Location
Chr.8: 13083361-13604606 on Build GRCh38
Transcripts
NM_001164271.1 NM_001316668.1 NM_006094.4 NM_182643.2 XM_005273374.1 XM_017012951.1 XM_017012952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7124696 c.3639A>G p.A1213A Chr.8: 13092713-13092713 on Build GRCh38
4993053 c.3697C>T p.L1233F Chr.8: 13092655-13092655 on Build GRCh38
6200601 c.3643G>A p.V1215I Chr.8: 13092709-13092709 on Build GRCh38
5506267 c.3642C>T p.A1214A Chr.8: 13092710-13092710 on Build GRCh38
1454813 c.3679G>A p.V1227M Chr.8: 13092673-13092673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144644763 Chr.8: 13092662-13092662 on Build GRCh38
rs775263028 Chr.8: 13092576-13092576 on Build GRCh38
rs370085276 Chr.8: 13092726-13092726 on Build GRCh38
rs142586910 Chr.8: 13092725-13092725 on Build GRCh38
rs768450831 Chr.8: 13092561-13092561 on Build GRCh38
rs774751541 Chr.8: 13092607-13092607 on Build GRCh38
rs907762073 Chr.8: 13092567-13092567 on Build GRCh38
rs370207065 Chr.8: 13092709-13092709 on Build GRCh38
rs201829040 Chr.8: 13092724-13092724 on Build GRCh38
rs34094992 Chr.8: 13092580-13092581 on Build GRCh38
rs774396057 Chr.8: 13092730-13092730 on Build GRCh38
rs779798978 Chr.8: 13092608-13092608 on Build GRCh38
rs200392347 Chr.8: 13092657-13092657 on Build GRCh38
rs201136617 Chr.8: 13092673-13092673 on Build GRCh38
rs766679826 Chr.8: 13092579-13092579 on Build GRCh38
rs769550422 Chr.8: 13092576-13092577 on Build GRCh38
rs201419347 Chr.8: 13092600-13092600 on Build GRCh38
rs554332281 Chr.8: 13092549-13092549 on Build GRCh38
rs746762491 Chr.8: 13092619-13092619 on Build GRCh38
rs983089854 Chr.8: 13092680-13092680 on Build GRCh38
rs138109577 Chr.8: 13092710-13092710 on Build GRCh38
rs764640253 Chr.8: 13092674-13092674 on Build GRCh38
rs200677979 Chr.8: 13092708-13092708 on Build GRCh38
rs769938928 Chr.8: 13092646-13092646 on Build GRCh38
rs376996403 Chr.8: 13092722-13092722 on Build GRCh38
rs775739963 Chr.8: 13092552-13092552 on Build GRCh38
rs1002011037 Chr.8: 13092584-13092584 on Build GRCh38
rs199784591 Chr.8: 13092599-13092599 on Build GRCh38
rs369659209 Chr.8: 13092656-13092656 on Build GRCh38
rs372435084 Chr.8: 13092564-13092564 on Build GRCh38
rs762299647 Chr.8: 13092637-13092637 on Build GRCh38
rs759769870 Chr.8: 13092663-13092663 on Build GRCh38
rs781236191 Chr.8: 13092628-13092628 on Build GRCh38
rs767895731 Chr.8: 13092665-13092665 on Build GRCh38
rs751261328 Chr.8: 13092698-13092698 on Build GRCh38
rs199509552 Chr.8: 13092580-13092580 on Build GRCh38
rs776762106 Chr.8: 13092562-13092562 on Build GRCh38
rs777804142 Chr.8: 13092711-13092711 on Build GRCh38
rs974465697 Chr.8: 13092523-13092523 on Build GRCh38
rs372133295 Chr.8: 13092596-13092596 on Build GRCh38
rs974163045 Chr.8: 13092570-13092570 on Build GRCh38
rs377046871 Chr.8: 13092707-13092707 on Build GRCh38
rs201392685 Chr.8: 13092581-13092581 on Build GRCh38
rs891355393 Chr.8: 13092611-13092611 on Build GRCh38
rs142779460 Chr.8: 13092685-13092685 on Build GRCh38
rs778396952 Chr.8: 13092602-13092602 on Build GRCh38
rs148049202 Chr.8: 13092531-13092531 on Build GRCh38
rs1030245940 Chr.8: 13092534-13092534 on Build GRCh38
rs748101950 Chr.8: 13092638-13092638 on Build GRCh38
rs1044094 Chr.8: 13092727-13092727 on Build GRCh38
rs763099740 Chr.8: 13092578-13092578 on Build GRCh38
rs991124482 Chr.8: 13092605-13092605 on Build GRCh38
rs770025144 Chr.8: 13092572-13092572 on Build GRCh38
rs1008375035 Chr.8: 13092635-13092635 on Build GRCh38
rs186299501 Chr.8: 13092548-13092548 on Build GRCh38
rs768598701 Chr.8: 13092626-13092626 on Build GRCh38
rs7829535 Chr.8: 13092601-13092601 on Build GRCh38
rs773154664 Chr.8: 13092647-13092647 on Build GRCh38
rs757976245 Chr.8: 13092683-13092683 on Build GRCh38
rs749871608 Chr.8: 13092677-13092677 on Build GRCh38
rs773502413 Chr.8: 13092573-13092573 on Build GRCh38

View Full Product Details