Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47161650-47161919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM2
Entrez Gene ID
805
Gene Name
calmodulin 2
Gene Aliases
CAMII, LQT15, PHKD, PHKD2, caM
UniGene
Hs.468442
Gene Chromosome Location
Chr.2: 47160082-47176936 on Build GRCh38
Transcripts
NM_001305624.1 NM_001305625.1 NM_001305626.1 NM_001743.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283910 c.290G>C p.G97A Chr.2: 47161854-47161854 on Build GRCh38
4545645 c.382G>A p.E128K Chr.2: 47161762-47161762 on Build GRCh38
6232984 c.360A>G p.E120E Chr.2: 47161784-47161784 on Build GRCh38
3581871 c.319C>T p.R107C Chr.2: 47161825-47161825 on Build GRCh38
4612613 c.417_418insCT p.E140fs*7 Chr.2: 47161726-47161727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374968950 Chr.2: 47161716-47161716 on Build GRCh38
rs199921656 Chr.2: 47161690-47161690 on Build GRCh38
rs398124648 Chr.2: 47161748-47161748 on Build GRCh38
rs200594401 Chr.2: 47161740-47161740 on Build GRCh38
rs751334551 Chr.2: 47161808-47161808 on Build GRCh38
rs546507069 Chr.2: 47161796-47161796 on Build GRCh38
rs761547095 Chr.2: 47161823-47161823 on Build GRCh38
rs775065505 Chr.2: 47161765-47161765 on Build GRCh38
rs542095802 Chr.2: 47161702-47161702 on Build GRCh38
rs750307914 Chr.2: 47161863-47161863 on Build GRCh38
rs751754595 Chr.2: 47161712-47161713 on Build GRCh38
rs2016682 Chr.2: 47161705-47161705 on Build GRCh38
rs11551437 Chr.2: 47161833-47161833 on Build GRCh38
rs201444040 Chr.2: 47161724-47161724 on Build GRCh38
rs773732456 Chr.2: 47161710-47161710 on Build GRCh38
rs17850324 Chr.2: 47161774-47161774 on Build GRCh38
rs398124649 Chr.2: 47161737-47161737 on Build GRCh38
rs756159706 Chr.2: 47161879-47161879 on Build GRCh38
rs34169936 Chr.2: 47161830-47161831 on Build GRCh38
rs398124647 Chr.2: 47161851-47161851 on Build GRCh38
rs779412690 Chr.2: 47161704-47161704 on Build GRCh38
rs3729955 Chr.2: 47161844-47161844 on Build GRCh38
rs372947059 Chr.2: 47161720-47161720 on Build GRCh38
rs1011415595 Chr.2: 47161853-47161853 on Build GRCh38
rs112882205 Chr.2: 47161877-47161878 on Build GRCh38
rs576033867 Chr.2: 47161706-47161706 on Build GRCh38
rs564738016 Chr.2: 47161841-47161841 on Build GRCh38
rs561747169 Chr.2: 47161875-47161876 on Build GRCh38
rs730882254 Chr.2: 47161857-47161857 on Build GRCh38
rs762591090 Chr.2: 47161790-47161790 on Build GRCh38
rs780167668 Chr.2: 47161885-47161885 on Build GRCh38
rs11551460 Chr.2: 47161843-47161843 on Build GRCh38
rs367611069 Chr.2: 47161701-47161701 on Build GRCh38
rs398124650 Chr.2: 47161744-47161744 on Build GRCh38

View Full Product Details