Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9917906-9918179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS7
Entrez Gene ID
8522
Gene Name
growth arrest specific 7
Gene Aliases
MLL/GAS7
UniGene
Hs.462214
Gene Chromosome Location
Chr.17: 9910609-10198551 on Build GRCh38
Transcripts
NM_001130831.1 NM_003644.2 NM_201432.1 NM_201433.1 XM_005256831.3 XM_005256832.3 XM_005256833.3 XM_005256834.3 XM_005256835.3 XM_011524044.2 XM_017025239.1 XM_017025240.1 XM_017025241.1 XM_017025242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6148289 c.863A>T p.Q288L Chr.17: 9918035-9918035 on Build GRCh38
6148288 c.1103A>T p.Q368L Chr.17: 9918035-9918035 on Build GRCh38
4604199 c.1247T>G p.V416G Chr.17: 9918071-9918071 on Build GRCh38
5350436 c.1102C>T p.Q368* Chr.17: 9918036-9918036 on Build GRCh38
4686460 c.1267C>A p.L423M Chr.17: 9918051-9918051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547644563 Chr.17: 9918137-9918137 on Build GRCh38
rs369028958 Chr.17: 9917983-9917983 on Build GRCh38
rs1009842180 Chr.17: 9917947-9917947 on Build GRCh38
rs754826416 Chr.17: 9917950-9917950 on Build GRCh38
rs759002085 Chr.17: 9918066-9918066 on Build GRCh38
rs753687681 Chr.17: 9918144-9918144 on Build GRCh38
rs775354136 Chr.17: 9918138-9918138 on Build GRCh38
rs773831004 Chr.17: 9918060-9918060 on Build GRCh38
rs371596444 Chr.17: 9918081-9918081 on Build GRCh38
rs562081230 Chr.17: 9918153-9918153 on Build GRCh38
rs550437632 Chr.17: 9918110-9918110 on Build GRCh38
rs369088042 Chr.17: 9917970-9917970 on Build GRCh38
rs747566446 Chr.17: 9918030-9918030 on Build GRCh38
rs62064494 Chr.17: 9918135-9918135 on Build GRCh38
rs779012858 Chr.17: 9918007-9918007 on Build GRCh38
rs376263450 Chr.17: 9918112-9918112 on Build GRCh38
rs780373030 Chr.17: 9918087-9918087 on Build GRCh38
rs886699877 Chr.17: 9918154-9918155 on Build GRCh38
rs376099685 Chr.17: 9917973-9917973 on Build GRCh38
rs1026592671 Chr.17: 9917941-9917941 on Build GRCh38
rs1050889116 Chr.17: 9917992-9917992 on Build GRCh38
rs2270122 Chr.17: 9918157-9918157 on Build GRCh38
rs145979040 Chr.17: 9918025-9918025 on Build GRCh38
rs367859092 Chr.17: 9918130-9918130 on Build GRCh38
rs183312776 Chr.17: 9917974-9917974 on Build GRCh38
rs748467211 Chr.17: 9918043-9918043 on Build GRCh38
rs368420647 Chr.17: 9917982-9917982 on Build GRCh38
rs749184881 Chr.17: 9918089-9918089 on Build GRCh38
rs369512800 Chr.17: 9917972-9917972 on Build GRCh38
rs1053669532 Chr.17: 9917933-9917933 on Build GRCh38
rs202125544 Chr.17: 9918083-9918083 on Build GRCh38
rs767477909 Chr.17: 9918070-9918070 on Build GRCh38
rs1003689165 Chr.17: 9918155-9918155 on Build GRCh38
rs11078822 Chr.17: 9917932-9917932 on Build GRCh38
rs750641887 Chr.17: 9918013-9918013 on Build GRCh38
rs1053614509 Chr.17: 9918131-9918131 on Build GRCh38
rs770124969 Chr.17: 9918105-9918105 on Build GRCh38
rs201170646 Chr.17: 9918080-9918080 on Build GRCh38
rs58204073 Chr.17: 9917990-9917990 on Build GRCh38
rs4791911 Chr.17: 9918150-9918150 on Build GRCh38
rs373995381 Chr.17: 9917964-9917964 on Build GRCh38
rs751215953 Chr.17: 9918077-9918078 on Build GRCh38
rs374485478 Chr.17: 9918091-9918091 on Build GRCh38
rs200175032 Chr.17: 9918059-9918059 on Build GRCh38
rs949507074 Chr.17: 9918094-9918094 on Build GRCh38
rs778171395 Chr.17: 9918107-9918107 on Build GRCh38
rs760440686 Chr.17: 9918077-9918077 on Build GRCh38
rs543813303 Chr.17: 9918136-9918136 on Build GRCh38
rs752569992 Chr.17: 9918071-9918071 on Build GRCh38
rs772128635 Chr.17: 9917966-9917966 on Build GRCh38
rs372193859 Chr.17: 9917961-9917961 on Build GRCh38
rs376131146 Chr.17: 9918029-9918029 on Build GRCh38
rs760627389 Chr.17: 9918140-9918140 on Build GRCh38
rs35312933 Chr.17: 9918055-9918055 on Build GRCh38
rs765901585 Chr.17: 9917977-9917977 on Build GRCh38
rs749554764 Chr.17: 9917956-9917956 on Build GRCh38
rs769308216 Chr.17: 9918031-9918031 on Build GRCh38
rs761474483 Chr.17: 9918147-9918147 on Build GRCh38
rs200968141 Chr.17: 9918001-9918001 on Build GRCh38
rs370226139 Chr.17: 9917965-9917965 on Build GRCh38
rs560176230 Chr.17: 9917940-9917940 on Build GRCh38
rs373878348 Chr.17: 9917979-9917979 on Build GRCh38
rs781768054 Chr.17: 9918104-9918104 on Build GRCh38
rs1020682150 Chr.17: 9917948-9917948 on Build GRCh38
rs565957216 Chr.17: 9918141-9918141 on Build GRCh38
rs900887666 Chr.17: 9918160-9918160 on Build GRCh38
rs772799704 Chr.17: 9918049-9918049 on Build GRCh38
rs568083945 Chr.17: 9918037-9918037 on Build GRCh38
rs752514969 Chr.17: 9918000-9918000 on Build GRCh38
rs374037924 Chr.17: 9918117-9918117 on Build GRCh38
rs755040901 Chr.17: 9918102-9918102 on Build GRCh38
rs372716070 Chr.17: 9917969-9917969 on Build GRCh38
rs780963567 Chr.17: 9917952-9917952 on Build GRCh38

View Full Product Details