Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37363147-37363416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKR1
Entrez Gene ID
284459
Gene Name
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases
ZNF875
UniGene
Hs.657027
Gene Chromosome Location
Chr.19: 37317900-37364455 on Build GRCh38
Transcripts
NM_181786.2 XM_017026672.1 XM_017026673.1 XM_017026674.1 XM_017026675.1 XM_017026676.1 XM_017026677.1 XM_017026678.1 XM_017026679.1 XM_017026680.1 XM_017026681.1 XM_017026682.1 XM_017026683.1 XM_017026684.1 XM_017026685.1 XM_017026686.1 XM_017026687.1 XM_017026688.1 XM_017026689.1 XM_017026690.1 XM_017026691.1 XM_017026692.1 XM_017026693.1 XM_017026694.1 XM_017026695.1 XM_017026696.1 XM_017026697.1 XM_017026698.1 XM_017026699.1 XM_017026700.1 XM_017026701.1 XM_017026702.1 XM_017026703.1 XM_017026704.1 XM_017026705.1 XM_017026707.1 XM_017026708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680673 c.1340T>A p.V447D Chr.19: 37363192-37363192 on Build GRCh38
6680671 c.1466G>A p.R489Q Chr.19: 37363318-37363318 on Build GRCh38
4923562 c.1431A>T p.P477P Chr.19: 37363283-37363283 on Build GRCh38
6680668 c.1596A>G p.R532R Chr.19: 37363391-37363391 on Build GRCh38
4690594 c.1476delG p.E494fs*19 Chr.19: 37363271-37363271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773180855 Chr.19: 37363336-37363336 on Build GRCh38
rs780521811 Chr.19: 37363311-37363311 on Build GRCh38
rs755034166 Chr.19: 37363390-37363390 on Build GRCh38
rs771050112 Chr.19: 37363342-37363342 on Build GRCh38
rs753356332 Chr.19: 37363285-37363285 on Build GRCh38
rs766387092 Chr.19: 37363283-37363283 on Build GRCh38
rs988666008 Chr.19: 37363290-37363290 on Build GRCh38
rs367884776 Chr.19: 37363352-37363352 on Build GRCh38
rs749379348 Chr.19: 37363256-37363256 on Build GRCh38
rs150628887 Chr.19: 37363386-37363386 on Build GRCh38
rs749339904 Chr.19: 37363316-37363316 on Build GRCh38
rs753375552 Chr.19: 37363181-37363181 on Build GRCh38
rs762646335 Chr.19: 37363364-37363364 on Build GRCh38
rs141902493 Chr.19: 37363371-37363371 on Build GRCh38
rs148655088 Chr.19: 37363349-37363349 on Build GRCh38
rs752330599 Chr.19: 37363304-37363304 on Build GRCh38
rs769889310 Chr.19: 37363218-37363218 on Build GRCh38
rs768712085 Chr.19: 37363317-37363317 on Build GRCh38
rs764766874 Chr.19: 37363353-37363353 on Build GRCh38
rs746100700 Chr.19: 37363355-37363356 on Build GRCh38
rs746336952 Chr.19: 37363212-37363212 on Build GRCh38
rs754628248 Chr.19: 37363185-37363185 on Build GRCh38
rs754497347 Chr.19: 37363294-37363294 on Build GRCh38
rs757585585 Chr.19: 37363205-37363205 on Build GRCh38
rs754327590 Chr.19: 37363389-37363389 on Build GRCh38
rs201417458 Chr.19: 37363328-37363328 on Build GRCh38
rs776575992 Chr.19: 37363345-37363345 on Build GRCh38
rs3745765 Chr.19: 37363333-37363333 on Build GRCh38
rs764086534 Chr.19: 37363187-37363188 on Build GRCh38
rs767378675 Chr.19: 37363271-37363271 on Build GRCh38
rs1050520680 Chr.19: 37363299-37363299 on Build GRCh38
rs757065327 Chr.19: 37363248-37363251 on Build GRCh38
rs781669442 Chr.19: 37363209-37363209 on Build GRCh38
rs760742788 Chr.19: 37363281-37363281 on Build GRCh38
rs751550901 Chr.19: 37363248-37363250 on Build GRCh38
rs942550230 Chr.19: 37363315-37363315 on Build GRCh38
rs960694686 Chr.19: 37363189-37363189 on Build GRCh38
rs746212508 Chr.19: 37363309-37363309 on Build GRCh38
rs778623168 Chr.19: 37363190-37363190 on Build GRCh38
rs774572333 Chr.19: 37363261-37363261 on Build GRCh38
rs375294602 Chr.19: 37363318-37363318 on Build GRCh38
rs768814758 Chr.19: 37363257-37363257 on Build GRCh38
rs150608212 Chr.19: 37363295-37363295 on Build GRCh38
rs144017676 Chr.19: 37363337-37363337 on Build GRCh38
rs765937686 Chr.19: 37363176-37363176 on Build GRCh38
rs752317648 Chr.19: 37363357-37363357 on Build GRCh38
rs988174718 Chr.19: 37363202-37363202 on Build GRCh38
rs775598633 Chr.19: 37363220-37363220 on Build GRCh38
rs201964801 Chr.19: 37363306-37363306 on Build GRCh38
rs141484624 Chr.19: 37363272-37363272 on Build GRCh38
rs756409153 Chr.19: 37363375-37363375 on Build GRCh38
rs138119894 Chr.19: 37363305-37363305 on Build GRCh38
rs761515101 Chr.19: 37363270-37363270 on Build GRCh38
rs755715246 Chr.19: 37363174-37363174 on Build GRCh38
rs551626575 Chr.19: 37363206-37363206 on Build GRCh38
rs747392346 Chr.19: 37363191-37363191 on Build GRCh38
rs756677165 Chr.19: 37363310-37363310 on Build GRCh38
rs781191343 Chr.19: 37363302-37363302 on Build GRCh38
rs868053386 Chr.19: 37363227-37363227 on Build GRCh38
rs757418769 Chr.19: 37363241-37363241 on Build GRCh38

View Full Product Details