Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 170283873-170284098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL1
Entrez Gene ID
28514
Gene Name
delta like canonical Notch ligand 1
Gene Aliases
DELTA1, DL1, Delta
UniGene
Hs.379912
Gene Chromosome Location
Chr.6: 170282200-170291075 on Build GRCh38
Transcripts
NM_005618.3 XM_005266934.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3623004 c.1307C>T p.S436L Chr.6: 170283972-170283972 on Build GRCh38
5112588 c.1288C>T p.R430C Chr.6: 170283991-170283991 on Build GRCh38
94132 c.1367G>T p.G456V Chr.6: 170283912-170283912 on Build GRCh38
5948813 c.1286G>A p.C429Y Chr.6: 170283993-170283993 on Build GRCh38
3777324 c.1390G>A p.D464N Chr.6: 170283889-170283889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762016479 Chr.6: 170283979-170283979 on Build GRCh38
rs148319460 Chr.6: 170283923-170283923 on Build GRCh38
rs372354902 Chr.6: 170283990-170283990 on Build GRCh38
rs376472547 Chr.6: 170284010-170284010 on Build GRCh38
rs143249553 Chr.6: 170283896-170283896 on Build GRCh38
rs926235895 Chr.6: 170283983-170283983 on Build GRCh38
rs41269633 Chr.6: 170283903-170283903 on Build GRCh38
rs374095083 Chr.6: 170283926-170283926 on Build GRCh38
rs749633512 Chr.6: 170284045-170284045 on Build GRCh38
rs374750063 Chr.6: 170283944-170283944 on Build GRCh38
rs201816034 Chr.6: 170284032-170284032 on Build GRCh38
rs573265046 Chr.6: 170284033-170284033 on Build GRCh38
rs369504757 Chr.6: 170283938-170283938 on Build GRCh38
rs770545429 Chr.6: 170283909-170283909 on Build GRCh38
rs762833797 Chr.6: 170284009-170284009 on Build GRCh38
rs752512403 Chr.6: 170284007-170284007 on Build GRCh38
rs773584103 Chr.6: 170283989-170283989 on Build GRCh38
rs546391656 Chr.6: 170284019-170284019 on Build GRCh38
rs868738986 Chr.6: 170284030-170284030 on Build GRCh38
rs780333054 Chr.6: 170284039-170284039 on Build GRCh38
rs16901311 Chr.6: 170283949-170283949 on Build GRCh38
rs61757615 Chr.6: 170283980-170283980 on Build GRCh38
rs763449437 Chr.6: 170283922-170283922 on Build GRCh38
rs150138823 Chr.6: 170283937-170283937 on Build GRCh38
rs12214318 Chr.6: 170283956-170283956 on Build GRCh38
rs752583724 Chr.6: 170283942-170283942 on Build GRCh38
rs140518054 Chr.6: 170283927-170283927 on Build GRCh38
rs748072021 Chr.6: 170283895-170283895 on Build GRCh38
rs777016922 Chr.6: 170283916-170283916 on Build GRCh38
rs764997017 Chr.6: 170283991-170283991 on Build GRCh38
rs572034237 Chr.6: 170284001-170284001 on Build GRCh38
rs761854965 Chr.6: 170283971-170283971 on Build GRCh38
rs765997935 Chr.6: 170284066-170284066 on Build GRCh38
rs540404589 Chr.6: 170284046-170284046 on Build GRCh38
rs368066905 Chr.6: 170283936-170283936 on Build GRCh38
rs199599888 Chr.6: 170283904-170283904 on Build GRCh38
rs774614063 Chr.6: 170283972-170283972 on Build GRCh38
rs780844790 Chr.6: 170283951-170283951 on Build GRCh38
rs1039024205 Chr.6: 170283897-170283897 on Build GRCh38
rs777329322 Chr.6: 170283898-170283898 on Build GRCh38
rs746261600 Chr.6: 170283917-170283917 on Build GRCh38
rs141144962 Chr.6: 170284052-170284052 on Build GRCh38
rs775661497 Chr.6: 170283955-170283955 on Build GRCh38
rs142808021 Chr.6: 170283950-170283950 on Build GRCh38
rs772284787 Chr.6: 170284056-170284056 on Build GRCh38
rs775917742 Chr.6: 170283968-170283968 on Build GRCh38
rs992329016 Chr.6: 170283998-170283998 on Build GRCh38
rs775873204 Chr.6: 170283920-170283920 on Build GRCh38
rs369260528 Chr.6: 170284065-170284065 on Build GRCh38
rs566580320 Chr.6: 170283918-170283918 on Build GRCh38
rs369678725 Chr.6: 170284038-170284038 on Build GRCh38
rs376384629 Chr.6: 170284055-170284055 on Build GRCh38

View Full Product Details