Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53497059-53497332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1D
Entrez Gene ID
776
Gene Name
calcium voltage-gated channel subunit alpha1 D
Gene Aliases
CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
UniGene
Hs.476358
Gene Chromosome Location
Chr.3: 53495049-53813151 on Build GRCh38
Transcripts
NM_000720.3 NM_001128839.2 NM_001128840.2 XM_005265448.3 XM_011534094.2 XM_011534096.2 XM_011534100.2 XM_017007137.1 XM_017007138.1 XM_017007139.1 XM_017007140.1 XM_017007141.1 XM_017007142.1 XM_017007143.1 XM_017007144.1 XM_017007145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2780686 c.100C>G p.L34V Chr.3: 53497184-53497184 on Build GRCh38
2780693 c.195T>C p.A65A Chr.3: 53497279-53497279 on Build GRCh38
5128781 c.244C>A p.Q82K Chr.3: 53497328-53497328 on Build GRCh38
4441238 c.166G>A p.A56T Chr.3: 53497250-53497250 on Build GRCh38
2780695 c.201A>G p.Q67Q Chr.3: 53497285-53497285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767957783 Chr.3: 53497267-53497267 on Build GRCh38
rs991468238 Chr.3: 53497139-53497139 on Build GRCh38
rs758400808 Chr.3: 53497298-53497298 on Build GRCh38
rs759586333 Chr.3: 53497112-53497112 on Build GRCh38
rs377363390 Chr.3: 53497216-53497216 on Build GRCh38
rs77610564 Chr.3: 53497127-53497127 on Build GRCh38
rs762132463 Chr.3: 53497253-53497253 on Build GRCh38
rs778510463 Chr.3: 53497174-53497174 on Build GRCh38
rs78820127 Chr.3: 53497126-53497126 on Build GRCh38
rs910544944 Chr.3: 53497110-53497110 on Build GRCh38
rs746863213 Chr.3: 53497213-53497213 on Build GRCh38
rs765082147 Chr.3: 53497115-53497116 on Build GRCh38
rs775933698 Chr.3: 53497116-53497116 on Build GRCh38
rs541931890 Chr.3: 53497121-53497121 on Build GRCh38
rs769710409 Chr.3: 53497231-53497231 on Build GRCh38
rs1005593744 Chr.3: 53497276-53497276 on Build GRCh38
rs764380630 Chr.3: 53497125-53497125 on Build GRCh38
rs982022592 Chr.3: 53497203-53497203 on Build GRCh38
rs777777778 Chr.3: 53497116-53497117 on Build GRCh38
rs935163999 Chr.3: 53497233-53497233 on Build GRCh38
rs753384864 Chr.3: 53497272-53497272 on Build GRCh38
rs539335706 Chr.3: 53497217-53497217 on Build GRCh38
rs770301253 Chr.3: 53497115-53497115 on Build GRCh38
rs760371924 Chr.3: 53497137-53497137 on Build GRCh38
rs764773584 Chr.3: 53497294-53497294 on Build GRCh38
rs775557387 Chr.3: 53497240-53497240 on Build GRCh38
rs781312928 Chr.3: 53497218-53497218 on Build GRCh38
rs752623993 Chr.3: 53497197-53497197 on Build GRCh38
rs769218354 Chr.3: 53497246-53497246 on Build GRCh38
rs766003838 Chr.3: 53497141-53497141 on Build GRCh38
rs763383320 Chr.3: 53497242-53497242 on Build GRCh38
rs888119455 Chr.3: 53497232-53497232 on Build GRCh38
rs774682333 Chr.3: 53497252-53497252 on Build GRCh38
rs140749530 Chr.3: 53497306-53497306 on Build GRCh38
rs371735256 Chr.3: 53497109-53497109 on Build GRCh38
rs575300482 Chr.3: 53497289-53497289 on Build GRCh38
rs754407641 Chr.3: 53497146-53497146 on Build GRCh38
rs917022424 Chr.3: 53497155-53497155 on Build GRCh38
rs543405396 Chr.3: 53497116-53497116 on Build GRCh38
rs746130121 Chr.3: 53497230-53497230 on Build GRCh38
rs758415159 Chr.3: 53497212-53497212 on Build GRCh38
rs752325493 Chr.3: 53497297-53497297 on Build GRCh38
rs751925700 Chr.3: 53497128-53497128 on Build GRCh38
rs759240348 Chr.3: 53497284-53497284 on Build GRCh38
rs150067408 Chr.3: 53497156-53497156 on Build GRCh38
rs192959416 Chr.3: 53497144-53497144 on Build GRCh38

View Full Product Details