Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143433336-143433603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
-
UniGene
Hs.618456
Gene Chromosome Location
Chr.4: 143336830-143474565 on Build GRCh38
Transcripts
NM_002039.3 NM_207123.2 XM_006714167.3 XM_006714168.3 XM_017007966.1 XM_017007967.1 XM_017007968.1 XM_017007969.1 XM_017007970.1 XM_017007971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051589 c.447C>A p.T149T Chr.4: 143433570-143433570 on Build GRCh38
7160013 c.475C>A p.P159T Chr.4: 143433598-143433598 on Build GRCh38
6668372 c.457T>C p.S153P Chr.4: 143433580-143433580 on Build GRCh38
1207747 c.470C>A p.T157N Chr.4: 143433593-143433593 on Build GRCh38
1207746 c.379C>T p.P127S Chr.4: 143433502-143433502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748212158 Chr.4: 143433504-143433504 on Build GRCh38
rs199870847 Chr.4: 143433526-143433526 on Build GRCh38
rs56219424 Chr.4: 143433392-143433392 on Build GRCh38
rs376330375 Chr.4: 143433547-143433547 on Build GRCh38
rs755393818 Chr.4: 143433459-143433459 on Build GRCh38
rs776589442 Chr.4: 143433578-143433578 on Build GRCh38
rs199684708 Chr.4: 143433506-143433506 on Build GRCh38
rs766587403 Chr.4: 143433455-143433455 on Build GRCh38
rs766476491 Chr.4: 143433576-143433576 on Build GRCh38
rs748332703 Chr.4: 143433557-143433559 on Build GRCh38
rs139449137 Chr.4: 143433422-143433422 on Build GRCh38
rs771186458 Chr.4: 143433531-143433531 on Build GRCh38
rs781328639 Chr.4: 143433449-143433449 on Build GRCh38
rs942272558 Chr.4: 143433399-143433399 on Build GRCh38
rs28925897 Chr.4: 143433391-143433391 on Build GRCh38
rs775106183 Chr.4: 143433544-143433544 on Build GRCh38
rs370770322 Chr.4: 143433553-143433553 on Build GRCh38
rs544394556 Chr.4: 143433577-143433577 on Build GRCh38
rs751987552 Chr.4: 143433458-143433458 on Build GRCh38
rs937620063 Chr.4: 143433462-143433462 on Build GRCh38
rs897964328 Chr.4: 143433451-143433451 on Build GRCh38
rs983519268 Chr.4: 143433381-143433381 on Build GRCh38
rs749410523 Chr.4: 143433527-143433527 on Build GRCh38
rs1036574268 Chr.4: 143433414-143433414 on Build GRCh38

View Full Product Details